Skip to Content
Merck
All Photos(3)

Key Documents

H7523

Sigma-Aldrich

HEPES

BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)

Synonym(s):

4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O4S
CAS Number:
Molecular Weight:
238.30
Beilstein:
883043
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Quality Level

Assay

≥99.5% (titration)

form

powder

impurities

Insoluble matter, passes filter test

ign. residue (900 °C)

≤0.1% (as SO4)

loss

≤1.0% loss on drying, 110°C

pH

5.0-6.5 (1 M in H2O)

useful pH range

6.8-8.2

pKa (25 °C)

7.5

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%

cation traces

Al: ≤0.0005%
As: ≤0.0001%
Ba: ≤0.0005%
Bi: ≤0.0005%
Ca: ≤0.001%
Cd: ≤0.0005%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Li: ≤0.0005%
Mg: ≤0.0005%
Mn: ≤0.0005%
Mo: ≤0.0005%
Na: ≤0.005%
Ni: ≤0.0005%
Pb: ≤0.0005%
Sr: ≤0.0005%
Zn: ≤0.0005%

absorption

≤0.035 at 280 in H2O at 1 M
≤0.045 at 260 in H2O at 1 M

application(s)

diagnostic assay manufacturing

SMILES string

OCCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)

InChI key

JKMHFZQWWAIEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

HEPES has been described as one of the best all-purpose buffers available for biological research. At biological pH, the molecule is zwitterionic, and is effective as a buffer at pH 6.8 to 8.2. HEPES has been used in a wide variety of applications, including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg.

Application

HEPES has been used:
  • As a component of homogenisation buffer used in protein sample preparation
  • To neutralise PRP (platelet-rich plasma) solution as part of PRP preparation
  • As a component in 2X PBS buffer, which is used in phosphate calcium transfection in cell line preparation
  • As a supplement in RPMI 1640 medium used to culture human umbilical vein endothelial cells (HUVEC) and in M199 complete medium for ex-vivo treatment of testicular tissue.

Other Notes

Easily compare specifications for HEPES products with the HEPES specification table.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The inhibitory effect of intracerebroventricularly injected interleukin 1? on testosterone secretion in the rat: role of steroidogenic acute regulatory protein.
Ogilvie KM, et al.
Biology of Reproduction, 60(2), 527-533 (1999)
Hsp60-induced tolerance in the rotifer Brachionus plicatilis exposed to multiple environmental contaminants.
Wheelock CE, et al.
Archives of Environmental Contamination and Toxicology, 36(3), 281-287 (1999)
Intervertebral disc regeneration in an ex vivo culture system using mesenchymal stem cells and platelet-rich plasma.
Chen WH, et al.
Biomaterials, 30(29), 5523-5533 (2009)
In vitro tubulogenesis of endothelial cells by relaxation of the coupling extracellular matrix-cytoskeleton.
Deroanne CF, et al.
Cardiovascular Research, 49(3), 647-658 (2001)
Assessing the corrosion of biodegradable magnesium implants: a critical review of current methodologies and their limitations.
Kirkland NT, et al.
Acta Biomaterialia, 8(3), 925-936 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service