Skip to Content
Merck
All Photos(3)

Key Documents

G7528

Sigma-Aldrich

D-(+)-Glucose

≥99.5% (GC), BioXtra

Synonym(s):

Aldohexose, Glucopyranose, Dextrose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724615
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

biological source

maize

Quality Level

product line

BioXtra

Assay

≥99.5% (GC)

form

powder

optical activity

[α]20/D +53±2°, 3 hr, c = 10 in H2O(lit.)

technique(s)

gas chromatography (GC): suitable

impurities

Insoluble matter, passes filter test

ign. residue

≤0.1% (as SO4)

loss

≤0.1% loss on drying

color

colorless

solubility

H2O: 1 M

anion traces

sulfate (SO42-): ≤0.005%

cation traces

Al: ≤0.0005%
As: ≤0.0001%
Ba: ≤0.0005%
Bi: ≤0.0005%
Ca: ≤0.001%
Cd: ≤0.0005%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Li: ≤0.0005%
Mg: ≤0.0005%
Mn: ≤0.0005%
Mo: ≤0.0005%
Na: ≤0.005%
Ni: ≤0.0005%
Pb: ≤0.0005%
Sr: ≤0.0005%
Zn: ≤0.0005%

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Gene Information

human ... PYGM(5837)

Looking for similar products? Visit Product Comparison Guide

General description

D-Glucose, a monosaccharide carbohydrate also referred to as dextrose or aldohexose, is a vital component in both biochemical research and cell culture studies. Its high purity makes it an excellent source of energy for cells and an important component in metabolic studies.Ideal for use in cellular energy metabolism, fermentation, and other biochemical research applications, our D-Glucose is of high quality and cost-effective. Whether you′re studying the role of glucose in cell cultures or investigating its effect on cellular energy, our D-Glucose is the perfect choice as an ideal solution.

Application

D-(+)-Glucose has been used:
  • as a component of Tris-glucose-EDTA (TGE) buffer for the preparation of bacterial DNA
  • in the preparation of peptide dilution buffer (PDB)
  • in the preparation of M17 broth medium
  • in the preparation of phosphate-buffered saline (PBS) buffer, used in phosphate calcium transfection

Features and Benefits

From Saccharomyces cerevisiae, ≥ 99.5 % (GC), BioXtra, Suitable for Cell Culture and Insect Cell Culture

Preparation Note

The material may produce a yellowish solution if prepared at a higher concentration than 2 g + 15mL.

Other Notes

  • To gain a comprehensive understanding of our diverse offerings of monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
  • For more information or to make an inquiry about this product, please click here to fill out our online form. Our team will get back to you as soon as possible.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biomolecular screening of formylpeptide receptor ligands with a sensitive, quantitative, high-throughput flow cytometry platform
Edwards B S, et al.
Nature Protocols, 1(1), 59-59 (2006)
Carbohydrates
Medical Biochemistry (2017)
Prevalence of Staphylococcus aureus and staphylococcal enterotoxins in raw pork and uncooked smoked ham-a comparison of classical culturing detection and RFLP-PCR
Atanassova V, et al.
International Journal of Food Microbiology, 68(1-2), 105-113 (2001)
Biochemical parameters in toxicological studies in Africa: significance, principle of methods, data interpretation, and use in plant screenings
Toxicological Survey of African Medicinal Plants, 659-715 (2014)
Transfer of antibiotic resistance marker genes between lactic acid bacteria in model rumen and plant environments.
Toomey N, et al.
Applied and Environmental Microbiology, 75(10), 3146-3152 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service