Merck
  • Quantitative determination of 26 steroids in eggs from various species using liquid chromatography-triple quadrupole-mass spectrometry.
Quantitative determination of 26 steroids in eggs from various species using liquid chromatography-triple quadrupole-mass spectrometry.
Journal of chromatography. A (2014-07-16)
Xiaoxia Mi, Sicong Li, Yanhua Li, Kaiqiang Wang, Dan Zhu, Gang Chen
ABSTRACT
A method for analyzing 26 types of steroids in egg matrix was developed. The method used liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS) in electrospray ionization mode (ESI). The procedure involved extraction with acetonitrile and removal of phospholipids by zinc chloride (ZnCl₂) followed by SPE cleanup with a Plexa cartridge. The effect of ZnCl₂ on phospholipid removal was directly observed using the post column infusion procedure. The SPE washing and elution conditions were optimized using a shallow gradient procedure. The free and conjugated steroids forms were determined using enzyme hydrolysis. The developed method resulted in satisfactory precision (RSD≤15%), and the limits of quantification were between 0.05 and 25 ng/g depending on the steroid types. The recoveries ranged from 63.2% to 121.5%. Finally, the developed method was successfully applied to compare the steroids in eggs from different species (i.e., hen, duck, quail and pigeon eggs) or different raising system (i.e., normal vs. organic eggs). The steroids can be clearly clustered according to species and raising system. The hierarchical clustering analysis indicated similarity of the steroids among the species. The developed method is sensitive and useful for detection and quantification of steroids in eggs and can be used for residue control programs. In addition, the observed steroid content will provide a fundamental reference for food risk assessment analysis.
Product Description
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
USP
Estrone, United States Pharmacopeia (USP) Reference Standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Prednisolone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetic acid, analytical standard
Supelco
17α-Methyltestosterone, VETRANAL®, analytical standard
Supelco
4-Androstene-3,17-dione, VETRANAL®, analytical standard
Prednisolone for system suitability, European Pharmacopoeia (EP) Reference Standard
Prednisolone, European Pharmacopoeia (EP) Reference Standard
Estrone, European Pharmacopoeia (EP) Reference Standard
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Progesterone, VETRANAL®, analytical standard
Supelco
Corticosterone, VETRANAL®, analytical standard
Supelco
Prednisolone, VETRANAL®, analytical standard
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Diethylstilbestrol, VETRANAL®, analytical standard
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Prednisolone, ≥99%
Sigma-Aldrich
17α-Methyltestosterone, solid (photosensitive)
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Estrone, ≥99%