Skip to Content
Merck
All Photos(3)

Key Documents

P3972

Sigma-Aldrich

Progesterone

meets USP testing specifications

Synonym(s):

4-Pregnene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
Beilstein:
1915950
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

Assay

≥97%

form

solid

mp

128-132 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19H,4-11H2,1-3H3/t16-,17+,18-,19-,20-,21+/m0/s1

InChI key

RJKFOVLPORLFTN-LEKSSAKUSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Induces maturation and secretory activity of the uterine endothelium; suppresses ovulation. Progesterone is implicated in the etiology of breast cancer.

Other Notes

Steroid hormone produced by the corpus luteum.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nina Lisofsky et al.
NeuroImage, 118, 154-162 (2015-06-10)
Hippocampal volume has been shown to be sensitive to variations in estrogen and progesterone levels across rodents' estrous cycle. However, little is known about the covariation of hormone levels and brain structure in the course of the human menstrual cycle.
Koji Mizuhashi et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 34(8), 1387-1392 (2019-03-20)
The growth plate provides a substantial source of mesenchymal cells in the endosteal marrow space during endochondral ossification. The current model postulates that a group of chondrocytes in the hypertrophic zone can escape from apoptosis and transform into cells that
Rudolf Kaaks et al.
International journal of cancer, 134(8), 1947-1957 (2013-10-25)
Results from prospective studies on premenopausal serum hormone levels in relation to breast cancer risk have been inconclusive, especially with regard to tumor subtypes. Using a case-control study nested within the prospective European Prospective Investigation into Cancer and Nutrition (EPIC)
Danijela Konforte et al.
Clinical chemistry, 59(8), 1215-1227 (2013-05-03)
Pediatric endocrinopathies are commonly diagnosed and monitored by measuring hormones of the hypothalamic-pituitary-gonadal axis. Because growth and development can markedly influence normal circulating concentrations of fertility hormones, accurate reference intervals established on the basis of a healthy, nonhospitalized pediatric population
Cathrin Brisken
Nature reviews. Cancer, 13(6), 385-396 (2013-05-25)
Understanding the biology of the breast and how ovarian hormones impinge on it is key to rational new approaches in breast cancer prevention and therapy. Because of the success of selective oestrogen receptor modulators (SERMs), such as tamoxifen, and aromatase

Articles

Milli-Q® ultrapure water systems deliver water free of organic contaminants , including hormones present in a lab's tap water, making it suitable for the most sensitive LC-MS/MS analyses.

Milli-Q® ultrapure water systems deliver water free of organic contaminants , including hormones present in a lab's tap water, making it suitable for the most sensitive LC-MS/MS analyses.

Milli-Q® ultrapure water systems deliver water free of organic contaminants , including hormones present in a lab's tap water, making it suitable for the most sensitive LC-MS/MS analyses.

Milli-Q® ultrapure water systems deliver water free of organic contaminants , including hormones present in a lab's tap water, making it suitable for the most sensitive LC-MS/MS analyses.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service