Skip to Content
Merck
All Photos(1)

Key Documents

46665

Supelco

Progesterone

VETRANAL®, analytical standard

Synonym(s):

4-Pregnene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
Beilstein:
1915950
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

Assay

99.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

128-132 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19H,4-11H2,1-3H3/t16-,17+,18-,19-,20-,21+/m0/s1

InChI key

RJKFOVLPORLFTN-LEKSSAKUSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Induces maturation and secretory activity of the uterine endothelium; suppresses ovulation. Progesterone is implicated in the etiology of breast cancer.

Other Notes

Steroid hormone produced by the corpus luteum.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rudolf Kaaks et al.
International journal of cancer, 134(8), 1947-1957 (2013-10-25)
Results from prospective studies on premenopausal serum hormone levels in relation to breast cancer risk have been inconclusive, especially with regard to tumor subtypes. Using a case-control study nested within the prospective European Prospective Investigation into Cancer and Nutrition (EPIC)
Danijela Konforte et al.
Clinical chemistry, 59(8), 1215-1227 (2013-05-03)
Pediatric endocrinopathies are commonly diagnosed and monitored by measuring hormones of the hypothalamic-pituitary-gonadal axis. Because growth and development can markedly influence normal circulating concentrations of fertility hormones, accurate reference intervals established on the basis of a healthy, nonhospitalized pediatric population
Brett E Skolnick et al.
The New England journal of medicine, 371(26), 2467-2476 (2014-12-11)
Progesterone has been associated with robust positive effects in animal models of traumatic brain injury (TBI) and with clinical benefits in two phase 2 randomized, controlled trials. We investigated the efficacy and safety of progesterone in a large, prospective, phase
Xiaofei Sun et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(5), 2380-2389 (2014-01-29)
Leucine-rich repeat-containing G-protein-coupled receptor 5 (Lgr5) is expressed in many organs, including female reproductive organs, and is a stem cell marker in the stomach and intestinal epithelium, hair follicles, and ovarian surface epithelium. Despite ongoing studies, the definitive physiological functions
Cathrin Brisken
Nature reviews. Cancer, 13(6), 385-396 (2013-05-25)
Understanding the biology of the breast and how ovarian hormones impinge on it is key to rational new approaches in breast cancer prevention and therapy. Because of the success of selective oestrogen receptor modulators (SERMs), such as tamoxifen, and aromatase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service