Skip to Content
Merck
All Photos(3)

Key Documents

P0130

Sigma-Aldrich

Progesterone

≥99% (HPLC), powder, maturation of the uterine endothelium inducer

Synonym(s):

4-Pregnene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
Beilstein:
1915950
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Product Name

Progesterone, ≥99%

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥99%

form

powder

mp

128-132 °C (lit.)

solubility

chloroform: 50 mg/mL, clear, colorless to light yellow

originator

Abbott

shipped in

ambient

storage temp.

room temp

SMILES string

CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19H,4-11H2,1-3H3/t16-,17+,18-,19-,20-,21+/m0/s1

InChI key

RJKFOVLPORLFTN-LEKSSAKUSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Induces maturation and secretory activity of the uterine endothelium; suppresses ovulation.
Induces maturation and secretory activity of the uterine endothelium; suppresses ovulation. Progesterone is implicated in the etiology of breast cancer.

Features and Benefits

This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Other Notes

Steroid hormone produced by the corpus luteum.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alicia T Pate et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 317(2), R328-R336 (2019-05-30)
Nesfatin-1 is a peptide derived from the nucleobindin 2 (Nucb2) precursor protein that has been shown to exert potent effects on appetite and cardiovascular function in male animals. Sex hormones modulate the expression of Nucb2 in several species, including goldfish
Nina Lisofsky et al.
NeuroImage, 118, 154-162 (2015-06-10)
Hippocampal volume has been shown to be sensitive to variations in estrogen and progesterone levels across rodents' estrous cycle. However, little is known about the covariation of hormone levels and brain structure in the course of the human menstrual cycle.
Cathrin Brisken
Nature reviews. Cancer, 13(6), 385-396 (2013-05-25)
Understanding the biology of the breast and how ovarian hormones impinge on it is key to rational new approaches in breast cancer prevention and therapy. Because of the success of selective oestrogen receptor modulators (SERMs), such as tamoxifen, and aromatase
Rudolf Kaaks et al.
International journal of cancer, 134(8), 1947-1957 (2013-10-25)
Results from prospective studies on premenopausal serum hormone levels in relation to breast cancer risk have been inconclusive, especially with regard to tumor subtypes. Using a case-control study nested within the prospective European Prospective Investigation into Cancer and Nutrition (EPIC)
Rudolf Kaaks et al.
Journal of the National Cancer Institute, 97(10), 755-765 (2005-05-19)
Contrasting etiologic hypotheses about the role of endogenous sex steroids in breast cancer development among premenopausal women implicate ovarian androgen excess and progesterone deficiency, estrogen excess, estrogen and progesterone excess, and both an excess or lack of adrenal androgens (dehydroepiandrosterone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service