Merck
  • Profiling of thiol-containing compounds by stable isotope labeling double precursor ion scan mass spectrometry.
Profiling of thiol-containing compounds by stable isotope labeling double precursor ion scan mass spectrometry.
Analytical chemistry (2014-09-16)
Ping Liu, Yun-Qing Huang, Wen-Jing Cai, Bi-Feng Yuan, Yu-Qi Feng
ABSTRACT
Here we developed a novel strategy of isotope labeling in combination with high-performance liquid chromatography-double precursor ion scan mass spectrometry (IL-LC-DPIS-MS) analysis for nontargeted profiling of thiol-containing compounds. In this strategy, we synthesized a pair of isotope labeling reagents (ω-bromoacetonylquinolinium bromide, BQB; ω-bromoacetonylquinolinium-d7 bromide, BQB-d7) that contain a reactive group, an isotopically labeled moiety, and an ionizable group to selectively label thiol-containing compounds. The BQB and BQB-d7 labeled compounds can generate two characteristic product ions m/z 218 and 225, which contain an isotope tag and therefore were used for double precursor ion scans in mass spectrometry analysis. The peak pairs with characteristic mass differences can be readily extracted from the two precursor ion scan (PIS) spectra and assigned as potential thiol-containing candidates, which facilitates the identification of analytes. BQB and BQB-d7 labeled thiol-containing compounds can be clearly distinguished by generating two individual ion chromatograms. Thus, thiol-containing compounds from two samples labeled with different isotope reagents are ionized at the same time but recorded separately by mass spectrometry, offering good identification and accurate quantification by eliminating the MS response fluctuation and mutual interference from the two labeled samples. Using the IL-LC-DPIS-MS strategy, we profiled the thiol-containing compounds in beer and human urine, and 21 and 103 thiol candidates were discovered in beer and human urine, respectively. In addition, 9 and 17 thiol candidates in beer and human urine were successfully identified by further comparison with thiol standards or tandem mass spectrometry analysis. Taken together, the IL-LC-DPIS-MS method is demonstrated to be a promising strategy in the profiling of compounds with identical groups in metabolomics study.
Product Description
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycine, 99%, FCC
SAFC
Glycine
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glycine, European Pharmacopoeia (EP) Reference Standard
Glutathione, European Pharmacopoeia (EP) Reference Standard
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Supelco
Quinoline, analytical standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3-Mercapto-1-hexanol, AldrichCPR
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Quinoline, reagent grade, 98%