Recommended Products
form
liquid
SMILES string
CCCC(S)CCO
InChI
1S/C6H14OS/c1-2-3-6(8)4-5-7/h6-8H,2-5H2,1H3
InChI key
TYZFMFVWHZKYSE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chemical senses, 45(4), 303-311 (2020-03-20)
Pleasantness is a major dimension of odor percepts. While naturally encountered odors rely on mixtures of odorants, few studies have investigated the rules underlying the perceived pleasantness of odor mixtures. To address this issue, a set of 222 binary mixtures
Journal of agricultural and food chemistry, 65(38), 8426-8434 (2017-08-31)
Early leaf removal around the cluster zone is a common technique applied in cool climate viticulture, to regulate yield components and improve fruit quality. Despite the increasing amount of information on early leaf removal and its impact on total soluble
Journal of agricultural and food chemistry, 65(21), 4329-4340 (2017-05-10)
To study the influence of molecular structure on the sensory properties of mercaptoalkanols, homologous series of 1-mercaptoalkan-3-ols, 3-mercaptoalkan-1-ols, 2-mercaptoalkan-1-ols, 4-mercaptoalkan-2-ols, 3-mercapto-3-methylalkan-1-ols, 1-mercapto-2-methylalkan-3-ols, 3-mercapto-2-methylalkan-1-ols, and 2-ethyl-3-mercaptoalkan-1-ols were synthesized. Odor thresholds in air and odor qualities were determined, and the results obtained
Food microbiology, 87, 103358-103358 (2020-01-18)
Microbes influence the quality of agricultural commodities and contribute to their distinctive sensorial attributes. Increasingly studies have demonstrated not only differential geographic patterns in microbial communities and populations, but that these contribute to valuable regionally distinct agricultural product identities, the
Journal of chromatography. A, 1637, 461803-461803 (2021-01-01)
Gas chromatography-olfactometry-mass spectrometry (GC-O-MS) has been very useful in identifying aroma compounds from within the complex matrix of wine. Supplementary separation can be required to overcome co-elution of volatiles or other sensory-directed chromatographic strategies are needed, including multidimensional chromatography and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service