Skip to Content
Merck
All Photos(3)

Key Documents

G8790

Sigma-Aldrich

Glycine

≥98.5%, suitable for cell culture, non-animal source, meets EP, JP, USP testing specifications

Synonym(s):

Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Beilstein:
635782
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%

biological source

non-animal source

Quality Level

Agency

meets EP testing specifications
meets JP testing specifications
meets USP testing specifications

Assay

≥98.5%

form

powder

quality

meets EP, JP, USP testing specifications

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white

pH

5.9-6.4

pKa (25 °C)

(1) 2.35, (2) 9.60
2.35

mp

240 °C (dec.) (lit.)

solubility

H2O: 100 mg/mL

SMILES string

NCC(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Amino acid for use in cell culture media development applications and existing media formulations.

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard A Felix et al.
Brain structure & function, 220(5), 2639-2652 (2014-06-30)
The mammalian superior paraolivary nucleus (SPON) is a major source of GABAergic inhibition to neurons in the inferior colliculus (IC), a well-studied midbrain nucleus that is the site of convergence and integration for the majority ascending auditory pathways en route
Alexander Carvajal-González et al.
Brain : a journal of neurology, 137(Pt 8), 2178-2192 (2014-06-22)
The clinical associations of glycine receptor antibodies have not yet been described fully. We identified prospectively 52 antibody-positive patients and collated their clinical features, investigations and immunotherapy responses. Serum glycine receptor antibody endpoint titres ranged from 1:20 to 1:60 000.
Zoé Husson et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(28), 9418-9431 (2014-07-11)
The principal neurons of the cerebellar nuclei (CN), the sole output of the olivo-cerebellar system, receive a massive inhibitory input from Purkinje cells (PCs) of the cerebellar cortex. Morphological evidence suggests that CN principal cells are also contacted by inhibitory
Bethany L Johnson-Kerner et al.
Human molecular genetics, 24(5), 1420-1431 (2014-11-16)
Giant axonal neuropathy (GAN) is a progressive neurodegenerative disease caused by autosomal recessive mutations in the GAN gene resulting in a loss of a ubiquitously expressed protein, gigaxonin. Gene replacement therapy is a promising strategy for treatment of the disease;
Dmitrij A Lisak et al.
Biochimica et biophysica acta, 1853(9), 2104-2114 (2015-03-15)
Bax inhibitor-1 (BI-1) is an evolutionarily conserved pH-dependent Ca²⁺ leak channel in the endoplasmic reticulum and the founding member of a family of six highly hydrophobic mammalian proteins named transmembrane BAX inhibitor motif containing (TMBIM) 1-6 with BI-1 being TMBIM6.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service