Skip to Content
Merck
All Photos(4)

Key Documents

399388

Sigma-Aldrich

Formic acid

greener alternative

ACS reagent, ≥88%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOOH
CAS Number:
Molecular Weight:
46.03
Beilstein:
1209246
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

1.6 (vs air)

vapor pressure

44.8 mmHg ( 20 °C)

description

dilution test passes test

Assay

≥88%

form

liquid

autoignition temp.

1004 °F

expl. lim.

57 %

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.4% Acetic acid

evapn. residue

≤0.002%

color

APHA: ≤15

refractive index

n20/D 1.370 (lit.)

pH

2.2 (20 °C, 2.2 g/L)

bp

100-101 °C (lit.)

mp

8.2-8.4 °C (lit.)

solubility

water: miscible

density

1.22 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.002%
sulfite (SO32-): passes test

cation traces

Fe: ≤5 ppm
NH4+: ≤0.005%
heavy metals: ≤5 ppm (by ICP)

greener alternative category

storage temp.

room temp

SMILES string

OC=O

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)

InChI key

BDAGIHXWWSANSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalysis. Click here for more information.

Application

Reductant used in iron-catalyzed, greener reduction of aldehydes to alcohols .

Fast and selective iron-catalyzed transfer hydrogenations of aldehydes

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F

Flash Point(C)

48 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Kitamoto et al.
Laboratory investigation; a journal of technical methods and pathology, 57(2), 230-236 (1987-08-01)
The purpose of this study was to design a method by which immunoperoxidase staining can be applied to formalin-fixed, paraffin-embedded tissue sections to demonstrate amyloid deposits in cerebral and systemic amyloidotic tissues. We used anti-prion protein, anti-beta-protein, anti-amyloid A, and
Modification of Pd for formic acid decomposition by support grafted functional groups.
Jones S, et al.
Catalysis, Structure & Reactivity, 1(1), 19-24 (2015)
Carbon dioxide as a raw material: the synthesis of formic acid and its derivatives from CO2.
Leitner W.
Angewandte Chemie (International Edition in English), 34(20), 2207-2221 (1995)
Advances in Direct Formic Acid Fuel Cells: Fabrication of Efficient Ir/Pd Nanocatalysts for Formic Acid Electro-Oxidation.
Al-Akraa IM, et al.
International Journal of Electrochemical Science, 10, 3282-3290 (2015)
Raman spectral study of 'neat' formic acid and aqueous and organic solutions of formic acid.
Bartholomew RJ and Irish DE.
Journal of Raman Spectroscopy, 30(4), 325-334 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service