Merck
  • A critical role for Neurofascin in regulating action potential initiation through maintenance of the axon initial segment.
A critical role for Neurofascin in regulating action potential initiation through maintenance of the axon initial segment.
Neuron (2011-03-09)
Barbara Zonta, Anne Desmazieres, Arianna Rinaldi, Steven Tait, Diane L Sherman, Matthew F Nolan, Peter J Brophy
ABSTRACT
The axon initial segment (AIS) is critical for the initiation and propagation of action potentials. Assembly of the AIS requires interactions between scaffolding molecules and voltage-gated sodium channels, but the molecular mechanisms that stabilize the AIS are poorly understood. The neuronal isoform of Neurofascin, Nfasc186, clusters voltage-gated sodium channels at nodes of Ranvier in myelinated nerves: here, we investigate its role in AIS assembly and stabilization. Inactivation of the Nfasc gene in cerebellar Purkinje cells of adult mice causes rapid loss of Nfasc186 from the AIS but not from nodes of Ranvier. This causes AIS disintegration, impairment of motor learning and the abolition of the spontaneous tonic discharge typical of Purkinje cells. Nevertheless, action potentials with a modified waveform can still be evoked and basic motor abilities remain intact. We propose that Nfasc186 optimizes communication between mature neurons by anchoring the key elements of the adult AIS complex.
Sigma-Aldrich
StableCell MEM, With Earle′s salts, stable glutamine, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
StableCell MEM, Alpha Modification, With stable glutamine and sodium bicarbonate, without ribonucleosides and deoxyribonucleosides, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
StableCell MEM, Alpha Modification, With stable glutamine and sodium bicarbonate, with ribonucleosides and deoxyribonucleosides, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, with Earle′s salts and non-essential amino acids, without L-glutamine, phenol red and sodium bicarbonate, Modified, powder, suitable for cell culture
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Hanks′ salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Hanks′ salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, non-essential amino acids and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Hanks′ salts, L-glutamine and non-essential amino acids, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Hanks′ salts and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, Alpha Modification, with L-glutamine, ribonucleosides and deoxyribonucleosides, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, Joklik Modification, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, L-glutamine, and non-essential amino acids, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, Modified, with Earle′s salts and reduced NaHCO3. without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, 10 ×, With Hanks′ salts, without L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, Alpha Modification, with sodium bicarbonate, without L-glutamine, ribonucleosides and deoxyribonucleosides, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium Eagle, AutoMod, with Earle′s salts, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, Spinner Modification, with Earle′s salts and sodium bicarbonate, without calcium chloride and L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, Joklik Modification, with L-glutamine, without calcium chloride and sodium bicarbonate, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, HEPES Modification, with Earle′s salts, 25 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, Alpha Modification, with ribonucleosides, deoxyribonucleosides and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Hanks′ Balanced Salt solution, With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, Alpha Modification, with L-glutamine and sodium pyruvate, without ribonucleosides, deoxyribonucleosides and sodium bicarbonate, powder, suitable for cell culture