Skip to Content
Merck
All Photos(1)

Key Documents

H9394

Sigma-Aldrich

Hanks′ Balanced Salt solution

Modified, with sodium bicarbonate, without calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

HBSS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

NaHCO3: 0.35 g/L
glucose: 1.0 g/L (Dextro)
phenol red: 0.011 g/L

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Hanks′ Balanced Salt solution is recommended for suspension cultures or whenever cell clumping is a problem.

Application

Hanks′ Balanced Salt solution has been used:
  • as a component of the culture medium for culturing of mouse cortical slices
  • as a component of the dissociation buffer for incubation of human-induced pluripotent stem cells (hiPSCs) and hiPSC-derived cardiomyocytes (CMs)
  • for in situ perfusions of mice liver

Biochem/physiol Actions

Balanced salt solution in cell culture plays multiple roles such as, an irrigating, transporting and diluting fluid while maintaining intra- and extracellular osmotic balance, and provides cells with water and certain bulk inorganic ions essential for normal cell metabolism. It is also involved in providing a buffering system to maintain the medium within the physiological pH range (7.2-7.6) and provides the principal energy source for cell metabolism.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Norma Jäppinen et al.
Nature communications, 10(1), 281-281 (2019-01-19)
Macrophages serve multiple functions including immune regulation, morphogenesis, tissue homeostasis and healing reactions. The current paradigm holds that mammary gland macrophages first arise postnatally during the prepubertal period from the bone marrow-derived monocytes. Here we delineate the origins of tissue-resident
Adrian Westhaus et al.
Human gene therapy, 31(9-10), 575-589 (2020-02-01)
Adeno-associated virus (AAV) vectors are quickly becoming the vectors of choice for therapeutic gene delivery. To date, hundreds of natural isolates and bioengineered variants have been reported. While factors such as high production titer and low immunoreactivity are important to
Sunny Nigam et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(3), 670-684 (2016-01-23)
The performance of complex networks, like the brain, depends on how effectively their elements communicate. Despite the importance of communication, it is virtually unknown how information is transferred in local cortical networks, consisting of hundreds of closely spaced neurons. To
Pratyusha Mandal et al.
Viruses, 12(11) (2020-11-01)
Cytomegaloviruses all encode the viral inhibitor of caspase-8-induced apoptosis (vICA). After binding to this initiator caspase, vICA blocks caspase-8 proteolytic activity and ability to activate caspase-3 and/or caspase-7. In this manner, vICA has long been known to prevent apoptosis triggered
Margret Bessadottir et al.
PloS one, 7(12), e51296-e51296 (2012-12-12)
The lichen compound usnic acid (UA) is a lipophilic weak acid that acts as a proton shuttle and causes loss of mitochondrial inner membrane potential. In the current study we show that UA treatment induced the formation of autophagosomes in

Protocols

StableCell™ Trypsin solutions are designed to perform cell detachment as standard trypsin solutions do, without the need to aliquot, freeze, and thaw. This saves significant time before passaging.

Trypsin is commonly used for dissociating adherent cells from surfaces. A wide variety of trypsin solutions are available to meet your specific cell line requirements.

Trypsin is commonly used for dissociating adherent cells from surfaces. A wide variety of trypsin solutions are available to meet your specific cell line requirements.

Trypsin is commonly used for dissociating adherent cells from surfaces. A wide variety of trypsin solutions are available to meet your specific cell line requirements.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service