Skip to Content
Merck
All Photos(1)

Key Documents

M2279

Sigma-Aldrich

Minimum Essential Medium Eagle

With Earle′s salts and sodium bicarbonate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

EMEM, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

product name

Minimum Essential Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

phenol red: yes
sodium pyruvate: no
L-glutamine: no
HEPES: no
NaHCO3: yes
Earle’s salts (5% CO2): yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Minimum Essential Medium Eagle (MEM) is one of the most widely used synthetic cell culture media. It includes higher concentrations of amino acids so the medium more closely approximates the protein composition of cultured mammalian cells. MEM has been used for the cultivation of a wide variety of cells grown in monolayers and attached cell lines, such as fibroblasts, in the presence of fetal bovine serum (FBS), calf, or horse sera.

Application

Minimum Essential Medium Eagle has been used to culture the:
  • explants of aortic valve leaflets
  • human liver cancer cell line (HepG2) cells
  • human fetal brain progenitor-derived astrocyte cells (PDA)
  • breast cancer cell line (MCF-7)
  • human glioblastoma cell line (T98G)

Preparation Note

Supplement with 0.292 g/L L-glutamine.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaisa-Emilia Makkonen et al.
Journal of virology, 87(20), 11148-11159 (2013-08-09)
Baculoviruses are insect-specific viruses commonly found in nature. They are not able to replicate in mammalian cells but can transduce them when equipped with an appropriate mammalian cell active expression cassette. Although the viruses have been studied in several types
Emily R Trzeciak et al.
Cells, 11(6) (2022-03-26)
The cellular composition of the tumor microenvironment, including tumor, immune, stromal, and endothelial cells, significantly influences responses to cancer therapies. In this study, we analyzed the impact of oxidative stress, induced by cold atmospheric plasma (CAP), on tumor cells, T
Luciano A Masullo et al.
Nature communications, 9(1), 3281-3281 (2018-08-18)
The theoretically unlimited spatial resolution of fluorescence nanoscopy often comes at the expense of time, contrast and increased dose of energy for recording. Here, we developed MoNaLISA, for Molecular Nanoscale Live Imaging with Sectioning Ability, a nanoscope capable of imaging
Winston X Yan et al.
Nature communications, 8, 15058-15058 (2017-05-13)
Precisely measuring the location and frequency of DNA double-strand breaks (DSBs) along the genome is instrumental to understanding genomic fragility, but current methods are limited in versatility, sensitivity or practicality. Here we present Breaks Labeling In Situ and Sequencing (BLISS)
Anna Bodzon-Kulakowska et al.
Cytotechnology, 67(6), 1085-1091 (2014-05-08)
Desorption electrospray ionization may be used as a fast and convenient method for analysis and identification of lipids in the cell culture. Oxidative stress, which usually involves changes in lipids, was used as a model of pathology to show the

Articles

Calcium is an ionically stable divalent cation with important beneficial and toxic properties in cell culture. It is a component of a wide range of cell culture media.

Calcium is an ionically stable divalent cation with important beneficial and toxic properties in cell culture. It is a component of a wide range of cell culture media.

Calcium is an ionically stable divalent cation with important beneficial and toxic properties in cell culture. It is a component of a wide range of cell culture media.

Calcium is an ionically stable divalent cation with important beneficial and toxic properties in cell culture. It is a component of a wide range of cell culture media.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service