Skip to Content
Merck
All Photos(1)

Key Documents

H9269

Sigma-Aldrich

Hanks′ Balanced Salt solution

With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

HBSS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

NaHCO3: 0.35 g/L
phenol red: 0.011 g/L
glucose: 1.0 g/L (Dextro)

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Formulation: click here.

Application

Hanks′ balanced salt solution has been used:
  • in starvation experiments for mammalian cells
  • to collect and maintain brain pieces for primary glial cell culture
  • to wash femoral tissue and bone marrow (BM) to culture mesenchymal stem cells

Biochem/physiol Actions

Balanced salt solution (BSS) in cell culture is multifaceted and has various functions. It serves as an irrigating, transporting, and diluting fluid while maintaining intra- and extracellular osmotic balance. BSS provides cells with water and certain bulk inorganic ions essential for normal cell metabolism. It combines with carbohydrates, such as glucose to act as the principal energy source for cell metabolism. BSS also provides a buffering system to maintain the medium within the physiological pH range (7.2-7.6).

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonya Nassari et al.
Scientific reports, 7(1), 17279-17279 (2017-12-10)
Connective tissues (CT) support and connect organs together. Understanding the formation of CT is important, as CT deregulation leads to fibrosis. The identification of CT specific markers has contributed to a better understanding of CT function during development. In developing
Ruyi Zhang et al.
Liver international : official journal of the International Association for the Study of the Liver, 41(1), 206-219 (2020-10-22)
Mature transfer RNAs (tRNA) charged with amino acids decode mRNA to synthesize proteins. Dysregulation of translational machineries has a fundamental impact on cancer biology. This study aims to map the tRNAome landscape in liver cancer patients and to explore potential
Muzaffar A Khan et al.
Toxins, 12(10) (2020-10-08)
Venomous snakes are important subjects of study in evolution, ecology, and biomedicine. Many venomous snakes have alpha-neurotoxins (α-neurotoxins) in their venom. These toxins bind the alpha-1 nicotinic acetylcholine receptor (nAChR) at the neuromuscular junction, causing paralysis and asphyxia. Several venomous
Xavier Taylor et al.
Journal of neuroinflammation, 17(1), 223-223 (2020-07-28)
Cerebral amyloid angiopathy (CAA) is typified by the cerebrovascular deposition of amyloid. The mechanisms underlying the contribution of CAA to neurodegeneration are not currently understood. Although CAA is highly associated with the accumulation of amyloid beta (Aβ), other amyloids are
Sarah A Wexler et al.
British journal of haematology, 121(2), 368-374 (2003-04-16)
In postnatal life, mesenchymal stem cells (MSC) self-replicate, proliferate and differentiate into mesenchymal tissues, including bone, fat, tendon, muscle and bone marrow (BM) stroma. Possible clinical applications for MSC in stem cell transplantation have been proposed. We have evaluated the

Protocols

3D Spheroid BME Cell Invasion Assay utilizes specialized plates and ECM for cell aggregation and spheroid formation in vitro.

3D Spheroid BME Cell Invasion Assay utilizes specialized plates and ECM for cell aggregation and spheroid formation in vitro.

3D Spheroid BME Cell Invasion Assay utilizes specialized plates and ECM for cell aggregation and spheroid formation in vitro.

3D Spheroid BME Cell Invasion Assay utilizes specialized plates and ECM for cell aggregation and spheroid formation in vitro.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service