Skip to Content
Merck
All Photos(1)

Key Documents

H8264

Sigma-Aldrich

Hanks′ Balanced Salt solution

Modified, with sodium bicarbonate, without phenol red, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

HBSS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

phenol red: no
glucose: 1.0 g/L (Dextro)
NaHCO3: 0.35 g/L

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Hanks’ Balanced Salt solution (HBSS) is a buffer that mimics biological medium and stabilizes cells. It is useful in leucocyte labeling studies. HBSS is an effective preservative for periodontal ligament cells. HBSS lacks essential inorganic ions and is useful in starvation studies.

Application

Calcium and magnesium are included in this formulation and support cell adhesion. Used to minimize cell attachment or aggregation.


Hanks’ Balanced Salt solution has been used:

  • as a rinse solution for hepatocytes
  • for hippocampi dissection
  • in corrosion testing of dog-bone specimens
  • for starvation studies in breast cancer cell line
  • as a component of dissociation solution for dorsal root ganglion
  • as a chamber solution for the patch-clamp apparatus

Other Notes

Phenol red has been shown to interfere with the growth of some cells at cloning densities. Use this medium when working with stem cells or when growing cells at low densities.

supplement

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hank s balanced salt solution: an alternative resuspension medium to label autologous leukocytes. Experience in inflammatory bowel disease
Martin-Comin J, et al.
Brazilian Archives of Biology and Technology, 45, 39-44 (2002)
Online oxygen monitoring using integrated inkjet-printed sensors in a liver-on-a-chip system
Moya A, et al.
Lab on a chip, 18, 2023-2035 (2018)
Vladimir Benes et al.
Scientific reports, 5, 11590-11590 (2015-06-26)
microRNAs are an abundant class of small non-coding RNAs that control gene expression post-transcriptionally. Importantly, microRNA activity participates in the regulation of cellular processes and is a potentially valuable source of biomarkers in the diagnosis and prognosis of human diseases.
Emmet Galvin et al.
Medical & biological engineering & computing, 55(8), 1261-1275 (2016-10-28)
Magnesium stents are a promising candidate in the emerging field of absorbable metallic stents (AMSs). In this study, the mechanical and corrosion performance of dog-bone specimens and a specific stent design of a magnesium alloy, WE43, are assessed experimentally in
Pauline Latzer et al.
Frontiers in cellular neuroscience, 13, 113-113 (2019-04-12)
A hallmark of glioblastoma multiforme (GBM) is neoangiogenesis, mediated by the overexpression of vascular endothelial growth factor (VEGF). Anti-VEGF antibodies, like bevacizumab, prolong progression-free survival in GBM, however, this treatment has been reported to be associated with a decline in

Related Content

Products and support for mpox research, offering world-class reagents for immunoassay and molecular assay development.

Products and support for mpox research, offering world-class reagents for immunoassay and molecular assay development.

Products and support for mpox research, offering world-class reagents for immunoassay and molecular assay development.

Products and support for mpox research, offering world-class reagents for immunoassay and molecular assay development.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service