Skip to Content
Merck
All Photos(1)

Key Documents

48732

Supelco

Aroclor 1242 solution

certified reference material, 50 mg/kg in transformer oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 5 mL

concentration

50 mg/kg in transformer oil

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

InChI

1S/C12H6Cl4/c13-7-1-3-9(11(15)5-7)10-4-2-8(14)6-12(10)16/h1-6H

InChI key

QORAVNMWUNPXAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M B Voltura et al.
Archives of environmental contamination and toxicology, 52(2), 264-269 (2006-12-13)
Studies of the impact of environmental contaminants on reproduction have typically focused on effects on fertility and subsequent reproductive failure. Contaminants may also impact reproductive output or other aspects of life history through effects on resource acquisition or allocation. We
Young-Cheol Cho et al.
Journal of microbiology (Seoul, Korea), 43(2), 166-171 (2005-05-10)
In order to determine the effects of sulfate concentration on the anaerobic dechlorination of polychlorinated biphenyls, sediments spiked with Aroclor 1242 were made into slurries using media which had various sulfate concentrations ranging from 3 to 23 mM. The time
Izabela Kania-Korwel et al.
Journal of chromatography. A, 1214(1-2), 37-46 (2008-11-21)
A pressurized liquid extraction-based method for the simultaneous extraction and in situ clean-up of polychlorinated biphenyls (PCBs), hydroxylated (OH)-PCBs and methylsulfonyl (MeSO(2))-PCBs from small (<0.5 g) tissue samples was developed and validated. Extraction of a laboratory reference material with hexane-dichloromethane-methanol
Shin-ichiro Narita et al.
Environmental health perspectives, 115(1), 48-52 (2007-03-21)
Prevalence and morbidity of allergic diseases have increased over the last decades. Based on the recently recognized differences in asthma prevalence between the sexes, we have examined the effect of endogenous estrogens on a key element of the allergic response.
Tarek Saba et al.
Chemosphere, 82(9), 1321-1328 (2010-12-31)
Aroclor 1242 contains a high percentage of lightly chlorinated congeners, which makes it susceptible to congener profile alterations as a result of physical-chemical environmental weathering by water washing, evaporation, and volatilization. The analysis of the variability of congener profiles in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service