Skip to Content
Merck
All Photos(1)

Key Documents

48735

Supelco

Aroclor 1260 solution

certified reference material, 500 mg/kg in transformer oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 5 mL

concentration

500 mg/kg in transformer oil

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

InChI

1S/C12H4Cl6/c13-7-3-1-5(9(15)11(7)17)6-2-4-8(14)12(18)10(6)16/h1-4H

InChI key

BTAGRXWGMYTPBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bianca Kuipers et al.
Canadian journal of microbiology, 49(1), 9-14 (2003-04-04)
We investigated the microbial reductive dechlorination of both weathered (aged) and nonweathered (freshly added) Aroclor 1260 in aerobic soil from Resolution Island, Nunavut, Canada. Initial polychlorinated biphenyl (PCB) concentrations were 106 and 100 ppm, respectively. The aerobic soil samples were
Chunlei Li et al.
Huan jing ke xue= Huanjing kexue, 23(2), 120-122 (2002-06-07)
The dosage of TiO2 and its surface-loaded silver had notable effect on the photocatalytic degradation of Aroclor1260, under ultraviolet light. The dosage of TiO2 had no proportion to the degradation rate of Aroclor1260, but had an optimal value, which was
Melissa Faria et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 149(3), 281-288 (2008-08-30)
Contaminant related changes in behavioral, phase I and II metabolizing enzymes and pro-oxidant/antioxidant processes in the freshwater mussels Dreissena polymorpha exposed to metals and PCBs were assessed. Behavioral and biochemical responses including filtering rates, key phase I, II and antioxidant
Taeko Tsuneta et al.
Environmental research, 107(2), 185-193 (2008-03-25)
Microbially mediated reductive dechlorination has been advocated as the first part of a two-stage (anaerobic/aerobic) biotreatment process for polychlorinated biphenyls (PCBs) in sediments, and is generally viewed as a detoxication process. However, previous studies suggest that microbial dechlorination increases the
H Y Yeung et al.
Chemosphere, 52(9), 1659-1665 (2003-07-18)
The differential expression of cytochrome P4501A1 gene expression (CYP1A1) in liver, intestine and gill of juvenile tilapia following oral exposure to polychlorinated biphenyl (PCB) Aroclor-1254 or Aroclor-1260 spiked food were investigated. The fish was fed with 0.1 g/g of body

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service