Skip to Content
Merck
All Photos(2)

Key Documents

A16254

Sigma-Aldrich

2-Furyl methyl ketone

99%

Synonym(s):

2-Acetylfuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O2
CAS Number:
Molecular Weight:
110.11
Beilstein:
107909
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.5070 (lit.)

bp

67 °C/10 mmHg (lit.)

mp

26-28 °C (lit.)

density

1.098 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1ccco1

InChI

1S/C6H6O2/c1-5(7)6-3-2-4-8-6/h2-4H,1H3

InChI key

IEMMBWWQXVXBEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Acute hepatitis in subjects exposed to 2-acetylfuran and hydrazine].
G Corsi et al.
La Medicina del lavoro, 74(4), 284-290 (1983-07-01)
T Kawai et al.
International archives of occupational and environmental health, 63(3), 213-219 (1991-01-01)
The apparent amount of 2,5-hexanedione, a biomarker of n-hexane expsoure in occupational health, in the urine of both exposed and non-exposed subjects varied not only as a function of the pH at which the urine sample was hydrolyzed but also
T R Bailey et al.
Journal of medicinal chemistry, 37(24), 4177-4184 (1994-11-25)
As a probe of the 3-methylisoxazole portion of our broad-spectrum antipicornaviral series, a panel of 2-acetylfuran analogues was prepared as replacements for the 3-methylisoxazole ring. Comparison of the two series showed remarkable similarity in potency, spectrum of activity, logP, and
Yu Wang et al.
Journal of agricultural and food chemistry, 56(24), 11997-12001 (2008-12-19)
Sugar type is a major factor regulating the reaction rates and pathways in Maillard reaction. Ribose and glucose were used to compare their reactivities and pathways of 2-acetylfuran formation. A stable isotope labeling method was used to study their reactivity.
Thomas Magauer et al.
Organic letters, 13(20), 5584-5587 (2011-10-01)
A 9-step synthetic route to the complex carbohydrate methyl α-trioxacarcinoside B from 2-acetylfuran is described. Anomerically activated forms, including 1-phenylthio, 1-O-(4'-pentenyl), 1-fluoro, and 1-O-acetyl derivatives are also prepared.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service