Skip to Content
Merck
All Photos(2)

Key Documents

53407

Supelco

5-(Hydroxymethyl)furfural

analytical standard

Synonym(s):

5-Hydroxymethyl-2-furaldehyde, 5-(Hydroxymethyl)furfural, 5-Hydroxymethyl-2-furancarboxaldehyde, HMF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
Beilstein:
110889
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC/HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.562 (lit.)

bp

114-116 °C/1 mmHg (lit.)

mp

28-34 °C (lit.)
28-34 °C (lit.)

density

1.243 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

[H]C(=O)c1ccc(CO)o1

InChI

1S/C6H6O3/c7-3-5-1-2-6(4-8)9-5/h1-3,8H,4H2

InChI key

NOEGNKMFWQHSLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-(Hydroxymethyl)furfural is an organic compound, widely spread in food products and is formed in sugar containing food when exposed to heat. It is widely used as an indicator of quality in food products. It is also employed to indicate the adulteration of food products with acid converted invert syrups.

Application

5-(Hydroxymethyl)furfural has been used as a standard in the high performance liquid chromatographic (HPLC) analysis for the determination of 5-(hydroxymethyl)furfural content in co-hydrolysates and fermentation broth of lignocellulosic biomass for the lipid accumulation in M. isabellina type of microorganisms. 5-(Hydroxymethyl)furfural may be used as an analytical reference standard for the quantification of the analyte in vinegar and fruit juice samples using chromatography techniques.
5-(Hydroxymethyl)furfural has been used as standard in the high performance liquid chromatographic (HPLC) analysis for the determination of 5-(hydroxymethyl)furfural content in co-hydrolysates and fermentation broth of lignocellulosic biomass for the lipid accumulation in M. isabellina type of microorganisms.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Co?hydrolysis of lignocellulosic biomass for microbial lipid accumulation
Ruan N, et al.
Biotechnology and Bioengineering, 110, 1039-1049 (2013)
Simultaneous determination of 5-hydroxymethylfurfural and patulin in apple juice by reversed-phase liquid chromatography
Gokmen V and Acar J
Journal of Chromatography A, 847(1-2), 69-74 (1999)
Determination of 5-hydroxymethylfurfural in vinegar samples by HPLC
Theobald A, et al.
Journal of Agricultural and Food Chemistry, 46(5), 1850-1854 (1998)
Yin-Ying Lee et al.
Physical chemistry chemical physics : PCCP, 14(40), 13914-13917 (2012-09-20)
Mesoporous silica nanoparticles functionalized with both sulfonic acid (HSO(3)) and ionic liquid (ILs) were synthesized and applied as effective and recyclable catalysts for generating 5-hydroxymethylfurfural (HMF) from fructose. For the first time a high HMF yield of 72.5% was achieved
Joanna Michalak et al.
International journal of environmental research and public health, 16(23) (2019-12-01)
Determining acrylamide (AA) content in foods using chromatographic methods is expensive and time-consuming. Therefore, there is a need to develop a simple, economical method for monitoring the content of acrylamide in foods. This study analysed whether there is a relationship

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service