Skip to Content
Merck
All Photos(1)

Key Documents

20258

Supelco

ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg

W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea

Synonym(s):

ORBO Supelpak 20 series Tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131900

material

W,W,W (specially treated) separators

Quality Level

Agency

NIOSH 2544,5506,5515,5800

product line

Amberlite
ORBO

composition

Bed A, 100 mg
Bed B, 50 mg

packaging

pkg of 50 ea

manufacturer/tradename

ORBO 43

technique(s)

active air sampling: suitable

O.D. × L

8 mm × 100 mm

matrix

XAD-2 (specially treated)

particle size

20-40 mesh

application(s)

air monitoring
environmental
industrial hygiene

Looking for similar products? Visit Product Comparison Guide

Application

ORBO 43 Supelpak-20 containing Amberlite XAD®- was suitable to collect gas-phase constituents during Aerodyne aerosol mass spectrometer (AMS).

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
ORBO is a trademark of Sigma-Aldrich Co. LLC
Supelpak is a trademark of Sigma-Aldrich Co. LLC
XAD is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naphthalene SOA: redox activity and naphthoquinone gas-particle partitioning.
McWhinney, R. D., S. Zhou, and J. P. D. Abbatt.
Atmospheric Chemistry and Physics, 13, 9731-9744 (2013)
Nikolaos G Tsiropoulos et al.
Analytica chimica acta, 573-574, 209-215 (2007-08-29)
A methodology is described for greenhouse air analysis by sampling fenhexamid, pyrimethanil, malathion, metalaxyl-M and myclobutanil in solid sorbents. Pesticides were determined by gas chromatography with NP Detector. The trapping efficiency of XAD-2, XAD-4, Supelpak-2, Florisil and C-18 at different
D Rochat et al.
Journal of chemical ecology, 17(11), 2127-2141 (1991-11-01)
Male American palm weevils (APWs),Rhynchophorus palmarum (L.) produced two sex-specific compounds, which were disclosed by volatile collections on Supelpak-2 and gas chromatography. One was a minor compound, not always detected. The major male-produced volatile was identified as (2E)-6-methyl-2-hepten-4-ol through coupled
F J Egea González et al.
Journal of AOAC International, 80(5), 1091-1097 (1997-11-05)
A method to sample and analyze chlorothalonil and dichlofluanid in greenhouse air was evaluated. Analysis was performed by gas chromatography with electron capture detection and gas chromatography-mass spectrometry. Solid sorbents such as Chromosorb 102, Porapak R, Supelpak-2, Amberlite XAD-2, Amberlite
A Orbo et al.
International journal of gynecological pathology : official journal of the International Society of Gynecological Pathologists, 22(2), 141-148 (2003-03-22)
Derangements in the tumor suppressor gene PTEN and the mismatch-repair genes, hMLH1, hMSH2, and hMSH6, have an important role in endometrial carcinogenesis. The purpose of this study was to assess immunohistochemically the pattern of protein expression for these genes in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service