20600-U
ORBO™ 1000 Precleaned Small PUF Plug, Replacement
O.D. × L 22 mm × 7.6 cm, pkg of 3 ea
Synonym(s):
Small PUF Cartridges
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12190000
NACRES:
NB.24
Recommended Products
material
polyurethane (foam)
Agency
ASTM® D4861,D4947
EPA IP-8,TO-10A
description
Replacement Plug
Small PUF
product line
ORBO™
feature
precleaned
packaging
pkg of 3 ea
manufacturer/tradename
ORBO™ 1000
technique(s)
active air sampling: suitable
O.D. × L
22 mm × 7.6 cm
matrix
PUF Polyurethane foam
application(s)
air monitoring
environmental
industrial hygiene
Looking for similar products? Visit Product Comparison Guide
General description
ORBO™-1000 small PUF cartridges are useful for ASTM D4861, EPA IP-8 and EPA TO-10A (Pesticides/PCBs); ASTM D4947 (Chlordane/Heptachlor).
Legal Information
ASTM is a registered trademark of American Society for Testing and Materials
ORBO is a trademark of Sigma-Aldrich Co. LLC
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service