Merck
  • Loss of expression of MLH1, MSH2, MSH6, and PTEN related to endometrial cancer in 68 patients with endometrial hyperplasia.
Loss of expression of MLH1, MSH2, MSH6, and PTEN related to endometrial cancer in 68 patients with endometrial hyperplasia.
International journal of gynecological pathology : official journal of the International Society of Gynecological Pathologists (2003-03-22)
A Orbo, M N Nilsen, M S Arnes, I Pettersen, K Larsen
ABSTRACT
Derangements in the tumor suppressor gene PTEN and the mismatch-repair genes, hMLH1, hMSH2, and hMSH6, have an important role in endometrial carcinogenesis. The purpose of this study was to assess immunohistochemically the pattern of protein expression for these genes in 68 patients with endometrial hyperplasia and to determine the relation of protein expression to cancer development or coexistence of cancers. Loss of expression of these genes also was evaluated as potential tumor markers for clinical use. PTEN and hMLH1 both showed loss of expression in 55% of specimens from 18 patients with subsequent or coexisting carcinoma. D&C specimens from 50 patients who did not develop cancer (10 patients underwent hysterectomy within 2 years; 40 had no hysterectomy; follow-up of 10-20 years), expressed protein at a much higher frequency (92% for PTEN and 98% for hMLH1). The parameter with the strongest independent relation to subsequent or coexisting carcinoma in a stepwise multiple logistic regression analysis was hMLH1. Evaluation of the investigated factors as prognostic markers for tumor development showed high specificity (92% for PTEN, 98% for MLH1) at the expense of sensitivity (56% for PTEN, 56% for MLH1). The results were compared with the results of the computerized image analysis algorithm, the D-score.
Product Description
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate