Skip to Content
Merck
All Photos(1)

Key Documents

U7128

Sigma-Aldrich

Uricase from Arthrobacter globiformis

lyophilized powder, 15-30 units/mg protein (biuret)

Synonym(s):

Urate oxidase, Urate:O2 oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

15-30 units/mg protein (biuret)

mol wt

tetramer ~32 kDa

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC3C(OC(C3O)N4C=CC(=O)NC4=O)CO)OCC1OC(C(C1O)O)N2C=CC(=O)NC2=O.[N+H4]

InChI

1S/C18H23N4O14P.H3N/c23-5-7-14(13(28)16(34-7)22-4-2-10(25)20-18(22)30)36-37(31,32)33-6-8-11(26)12(27)15(35-8)21-3-1-9(24)19-17(21)29;/h1-4,7-8,11-16,23,26-28H,5-6H2,(H,31,32)(H,19,24,29)(H,20,25,30);1H3

InChI key

VPXDHZMLJOJKOX-UHFFFAOYSA-N

General description

Research area: Cell Signalling. Uricase is made of four identical subunits and is a homotetrameric enzyme. It is found as a functional uricase in several organisms including, bacteria, archaea, and eukaryotes. Nonetheless, primates and humans lack a functional uricase enzyme.

Application

Uricase from Arthrobacter globiformis has been used for coating the electrodes in - biosensor fabrication for the detection of uric acid in urine samples. It has also been used for analysing the effect of Zn, Cu, Mg, and Mn ions on uricase.

Biochem/physiol Actions

Arthrobacter globiformis is a major producer of uricase or urate oxidase in poultry manure and litter. Uricase hydrolyzes uric acid and causes ammonia volatilization in poultry manure. In the majority of mammals, uricase converts uric acid to allantoin, a soluble excretion product. Humans lack uricase due to its loss during the evolution of hominids. This increases the risk of humans towards gout, kidney stones, hypertension, and diabetes due to increased circulating levels of uric acid.Uricase is an important enzyme that plays a role in the purine metabolic pathway.
Molecular weight: 4 × ~32 KDa (tetramer)
Isoelectric point: is 4.64
KM value (uric acid as substrate): ~ 0.01 mM
The optimum pH for activity: 8.5-9.5
The optimum pH for stability: 6.0-9.5
Zn and Cu were shown to inhibit the activity of uricase.

Unit Definition

One unit will convert 1.0 μmole of uric acid to allantoin per min at pH 9.0 at 25 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Therapeutic Uricase with Reduced Immunogenicity Risk and Improved Development Properties
Andrew C N, et al.
PLoS ONE, 11(12) (2016)
Development of a sandwich format, amperometric screen-printed uric acid biosensor for urine analysis.
Kanyong P, et al.
Analytical biochemistry, 428(1), 39-43 (2012)
Review of Uricase and Other Key Metabolites Produced by Microorganisms through Submerged Fermentation INTRODUCTION
Sandeep V C, et al.
Fluoropolymers 2. , 22(4) (2023)
Uric acid: a danger signal from the RNA world that may have a role in the epidemic of obesity, metabolic syndrome, and cardiorenal disease: evolutionary considerations.
Seminars in Nephrology, 31(5), 394-399 (2011)
Effect of minerals on activity of microbial uricase to reduce ammonia volatilization in poultry manure.
Kim WK and Patterson PH
Poultry Science, 82(2), 223-231 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service