Skip to Content
Merck
All Photos(1)

Key Documents

10110434001

Roche

Xanthine Oxidase (XOD)

from cow milk

Synonym(s):

XOD

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204

biological source

bovine milk

Quality Level

form

suspension

specific activity

~1 units/mg protein (At 25 °C with xanthine as the substrate.)

packaging

pkg of 1 mL (20 U)

manufacturer/tradename

Roche

optimum pH

8.5-9.0

shipped in

wet ice

storage temp.

2-8°C

Related Categories

General description

Xanthine Oxidase (XOD) is a metal flavoprotein. It has FAD, molybdenum and iron in the ratio 2:2:8.
Xanthine:oxygen oxidoreductase

Application

Xanthine Oxidase (XOD) has been used in the assessment of XOR-mediated NO production from NDHP and assessment of nitrite-derived NO in liver and purified XOR.
Xanthine Oxidase has been used to study tyrosine nitration.

Biochem/physiol Actions

Xanthine Oxidase (XOD) exhibits a broad substrate specificity including aldehydes, purines and pteridines. Furthermore, this enzyme reduces oxygen to generate superoxide, hydrogen peroxide and reactive oxygen species (ROS). It also reduces nitrite to yield reactive nitrogen species (RNS), such as peroxynitrite and nitric oxide. Owing to its ability to generate RNS and ROS, XOD might play an important role as an antimicrobial agent in the neonatal gut, thereby complementing endogenous enzyme of the intestinal epithelium.

Quality

Contaminants: <0.005% guanase, NP and uricase, each, <0.05% ADA, <0.05% alkaline phosphatase (4-nitrophenyl phosphate as the substrate)
Note: Chromatographically purified.

Sequence

XOD is a dimer. Each subunit contains 1 atom of molybdenum, 2 iron-sulfur centers (non-heme iron, ferredoxin-type) and 1 molecule of FAD.

Unit Definition

One unit (U) xanthine oxidase will produce 1 μmol of uric acid (E293nm = 12.2 mmol -1 x L x cm-1) from the oxidation of 1 μmol of xanthine in 1 min at 25 °C and pH8.5.

Physical form

Suspension in 3.2 M ammonium sulfate solution, 10 mM EDTA, pH approximately 8

Preparation Note

Activator: O2
Stabilizers: The substance is stabilized by the addition of EDTA; salicylate is not added.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The novel organic mononitrate NDHP attenuates hypertension and endothelial dysfunction in hypertensive rats.
Paulo LL, et al.
Redox Biology, 15, 182-191 (2018)
International Dairy Journal
R. Harrison.
Science, 16 (6), 546-554 (2006)
T Sawa et al.
The Journal of biological chemistry, 275(42), 32467-32474 (2000-07-25)
Peroxynitrite (ONOO(-)) is a potent nitrating and oxidizing agent that is formed by a rapid reaction of nitric oxide (NO) with superoxide anion (O(2)). It appears to be involved in the pathophysiology of many inflammatory and neurodegenerative diseases. It has
Methods of Enzymatic Analysis (2012)
XANTHINE OXIDASE. VI. INFLUENCE OF PH ON SUBSTRATE SPECIFICITY.
L GREENLEE et al.
The Journal of biological chemistry, 239, 1090-1095 (1964-04-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service