Skip to Content
Merck
All Photos(2)

Key Documents

U0880

Sigma-Aldrich

Uricase from Candida sp.

recombinant, expressed in E. coli, lyophilized powder, ≥2 units/mg solid

Synonym(s):

Urate: oxygen oxoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

form

lyophilized powder

specific activity

≥2 units/mg solid

mol wt

35 kDa by SDS-PAGE

storage temp.

−20°C

InChI

1S/C18H23N4O14P.H3N/c23-5-7-14(13(28)16(34-7)22-4-2-10(25)20-18(22)30)36-37(31,32)33-6-8-11(26)12(27)15(35-8)21-3-1-9(24)19-17(21)29;/h1-4,7-8,11-16,23,26-28H,5-6H2,(H,31,32)(H,19,24,29)(H,20,25,30);1H3

InChI key

VPXDHZMLJOJKOX-UHFFFAOYSA-N

General description

Uricase deficient mice develop uric acid nephropathy and diabetes insipidus which can be mitigated with recombinant uricase treatment.
Uricase is a homo-tetrameric protein with its active sites at the head-to-tail monomer-dimer interface.

Application

Uricase from Candida sp. has been used:
  • to eliminate urate in plasma samples and evaluate the contribution of urate to the total plasma antioxidant capacity
  • to intraperitoneally inject the mice to study the role of uric acid in fibrin extracellular traps (ET) formation
  • for elimination of uric acid in ferric reducing/antioxidative power (FRAP) assay to measure the total antioxidative capacity of human plasma sample; in uricase-based biosensors

Biochem/physiol Actions

Uricase participates in purine catabolism. It catalyzes the conversion of highly insoluble uric acid into 5-hydroxyisourate. Accumulation of uric acid causes liver/kidney damage or chronically causes gout. In mice, a mutation in the gene encoding uricase causes a sudden increase in uric acid. Mice, deficient in this gene, exhibit hyperuricemia, hyperuricosuria, and uric acid crystalline obstructive nephropathy.

Unit Definition

One unit will oxidize 1.0 μmole of uric acid to allantoin per minute at pH 8.5 at 25 °C.

Physical form

Powder; contains citrate and sucrose

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rocco Cancelliere et al.
Biosensors, 12(1) (2022-01-21)
The use of carbon nanomaterials (CNMs) in sensors and biosensor realization is one of the hottest topics today in analytical chemistry. In this work, a comparative in-depth study, exploiting different nanomaterial (MWNT-CO2H, -NH2, -OH and GNP) modified screen-printed electrodes (SPEs)
Darko Duplancic et al.
Molecules (Basel, Switzerland), 16(8), 7058-7068 (2011-08-19)
Determination of the relative contribution of uric acid level increases to the total measured antioxidative activity could be very useful for testing antioxidative products and their effect on human health. The aim of this report is to present a simple
Benoit Pugin et al.
Frontiers in microbiology, 13, 853735-853735 (2022-05-03)
Uricase catalyzes the conversion of uric acid into allantoin with concomitant reduction of molecular oxygen to hydrogen peroxide. In humans, uricase is not functional, thereby predisposing individuals to hyperuricemia, a metabolic disturbance associated with gout, chronic kidney disorders, and cardiovascular
Michael W Munks et al.
Blood, 116(24), 5191-5199 (2010-09-30)
It has been recognized for nearly 80 years that insoluble aluminum salts are good immunologic adjuvants and that they form long-lived nodules in vivo. Nodule formation has long been presumed to be central for adjuvant activity by providing an antigen
S J Kelly et al.
Journal of the American Society of Nephrology : JASN, 12(5), 1001-1009 (2001-04-24)
Uricase-deficient mice develop uric acid nephropathy, with high mortality rates before weaning. Urate excretion was quantitated and renal function was better defined in this study, to facilitate the use of these mice as a model for evaluating poly(ethylene glycol)-modified recombinant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service