Skip to Content
Merck
All Photos(1)

Key Documents

94310

Sigma-Aldrich

Uricase from Bacillus fastidiosus

lyophilized, ~9 U/mg

Synonym(s):

Urate oxidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.54

biological source

Bacillus sp. (B. fastidiosus)

Quality Level

form

powder

quality

lyophilized

specific activity

~9 U/mg

storage condition

dry at room temperature

concentration

90—100%

color

white to faint beige

solubility

5 mg/mL, clear, colorless to very faintly yellow (in enzyme dilution buffer)

application(s)

detection

storage temp.

−20°C

SMILES string

N.OCC1OC(C(O)C1OP(O)(=O)OCC2OC(C(O)C2O)N3C=CC(=O)NC3=O)N4C=CC(=O)NC4=O

InChI

1S/C18H23N4O14P.H3N/c23-5-7-14(13(28)16(34-7)22-4-2-10(25)20-18(22)30)36-37(31,32)33-6-8-11(26)12(27)15(35-8)21-3-1-9(24)19-17(21)29;/h1-4,7-8,11-16,23,26-28H,5-6H2,(H,31,32)(H,19,24,29)(H,20,25,30);1H3

InChI key

VPXDHZMLJOJKOX-UHFFFAOYSA-N

General description

Research area: Cell signaling
Uricase is present in many organisms, such as, archaea, bacteria, and eukaryotes. The Gut microbiome is the major source of uricase, a homotetrameric enzyme containing four identical 34 kDa subunits.

Application

Uricase may be used for the determination of uric acid in biological fluids. It may be helpful in the spectrophotometric determination of purines and purine nucleosides.

Biochem/physiol Actions

Uricase catalyzes the oxidation of uric acid to 5-hydroxyisourate, which subsequently forms (S)-allantoin. It is involved in purine metabolism, urate degradation and (S)-allantoin formation The absence of functional uricase in humans has significant physiological and pathophysiological implications. It has been associated with higher serum uric acid levels, leading to an increased risk of conditions related to uric acid crystallization, such as gout. Uricase has emerged as an important therapeutic target for the treatment of conditions related to hyperuricemia, such as gout.

Unit Definition

1 U corresponds to the amount of enzyme which oxidizes 1 μmole uric acid at pH 9.1 and 37 °C.

Other Notes

For the determination of uric acid in biological fluids; In the spectrophotometric determination of purines and purine nucleosides.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Resp. Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.H. Jensen et al.
Methods of Enzymatic Analysis, 7, 101-101 (1985)
Berna Dalkıran et al.
Mikrochimica acta, 187(11), 609-609 (2020-10-16)
Electropolymerization of thionine (TH) on multiwalled carbon nanotube (MWCNT)-modified glassy carbon electrodes (GCE) in ethaline deep eutectic solvent (DES) was carried out for the first time, to prepare poly(thionine) (PTH) films with different nanostructured morphologies. PTH films were formed on
A Therapeutic Uricase with Reduced Immunogenicity Risk and Improved Development Properties
Nyborg AC, et al.
PLoS ONE (2016)
Youssef M Roman
Journal of personalized medicine, 13(9) (2023-09-28)
Uric acid is the final product of purine metabolism and is converted to allantoin in most mammals via the uricase enzyme. The accumulation of loss of function mutations in the uricase gene rendered hominoids (apes and humans) to have higher
U. Naegele et al.
Methods of Enzymatic Analysis, 7, 140-140 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service