Skip to Content
Merck
All Photos(1)

Key Documents

S2263

Sigma-Aldrich

Sheep Serum

USA origin, sterile-filtered, suitable for cell culture

Synonym(s):

ovine sera, ovine serum, sheep sera

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

biological source

sheep

Quality Level

sterility

sterile-filtered

composition

hemoglobin, ≤50 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤100 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Application

Sheep serum is useful for the propagation of long-term cultures of erythrocytic stages of the Theileria uilenbergi intracellular protozoan parasite.

Preparation Note

Collected from adult animals.

Analysis Note

Endotoxin tested

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daigo Inoue et al.
PloS one, 6(5), e19713-e19713 (2011-05-24)
For the detection and sub-cellular (co)-localization of proteins in the context of the tissue or organism immunostaining in whole mount preparations or on sections is still the best approach. So far, each antibody required its own fixation and antigen retrieval
Rohola Hosseini et al.
Autophagy, 10(10), 1844-1857 (2014-08-16)
High-resolution imaging of autophagy has been used intensively in cell culture studies, but so far it has been difficult to visualize this process in detail in whole animal models. In this study we present a versatile method for high-resolution imaging
Mardi S Byerly et al.
PloS one, 8(11), e80781-e80781 (2013-12-07)
During development, prenatal and postnatal factors program homeostatic set points to regulate food intake and body weight in the adult. Combinations of genetic and environmental factors contribute to the development of neural circuitry that regulates whole-body energy homeostasis. Brain-derived neurotrophic
Ioana M Abbas et al.
Analytical and bioanalytical chemistry, 410(16), 3835-3846 (2018-04-19)
Mass spectrometry-based methods play a crucial role in the quantification of the main iron metabolism regulator hepcidin by singling out the bioactive 25-residue peptide from the other naturally occurring N-truncated isoforms (hepcidin-20, -22, -24), which seem to be inactive in
Ann Ng et al.
Developmental biology, 335(1), 208-215 (2009-09-08)
Glypican-3 (Gpc3) is a heparan sulfate proteoglycan (HSPG) expressed widely during vertebrate development. Loss-of-function mutations cause Simpson-Golabi-Behmel syndrome (SGBS), a rare and complex congenital overgrowth syndrome with a number of associated developmental abnormalities including congenital heart disease. We found that

Articles

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service