Skip to Content
Merck
All Photos(1)

Key Documents

RPOLT7-RO

Roche

T7 RNA Polymerase

from Escherichia coli BL 21/pAR 1219

Synonym(s):

polymerase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204

biological source

Escherichia coli (BL 21/pAR 1219)

Quality Level

Assay

100% (SDS-PAGE)

form

solution

specific activity

≥20 U/μL

mol wt

98  kDa (Single polypeptide chain)

packaging

pkg of 1,000 U (10881767001)
pkg of 5,000 U (10881775001)

manufacturer/tradename

Roche

technique(s)

Northern blotting: suitable
Southern blotting: suitable
hybridization: suitable

color

colorless

pH

7.9 (39 °F)

solubility

water: miscible

suitability

suitable for molecular biology

NCBI accession no.

UniProt accession no.

application(s)

life science and biopharma

foreign activity

Endonucleases 100 units, none detected
Nicking activity 100 units, none detected
RNase 100 units, none detected

storage temp.

−20°C

Gene Information

Escherichia coli ... T7p07(1261050)

General description

T7 RNA polymerase is commonly used to transcribe DNA which has been cloned into vectors which have two phage promoters in opposite orientation. RNA can be selectively synthesized from either strand of the insert DNA with different polymerases. Homogeneously labeled single-stranded RNA can be generated with this system. Transcripts can be non-radioactively labeled with biotin or DIG-11-UTP or radioactively labeled to high specific activity with [α-32P] or [α-35S]-labeled nucleotides.

Synthesis of hybridization probes: T7 RNA polymerase allows highly efficient production of homogeneously labeled RNA. This labeled RNA may be used as hybridization probes in Southern, northern, and dot blots, as well as in situ hybridizations.
Suitable labels:Transcripts can be nonradioactively labeled with biotin-16-UTP or DIG-11-UTP. They may also be radioactively labeled to high specific activity with [α-32P]- or [α-35S]-labeled nucleotides.

Specificity

Promotor specifity
T7 RNA polymerase is extremely promoter-specific and only transcribes bacteriophage T7 DNA or DNA cloned downstream of a T7 promoter. Although the T7 and T3 promoter sequences differ only by 3 bp, T7 RNA polymerase only transcribes DNA cloned downstream of its promoter.
Heat inactivation: Stop the reaction by adding 2 μl 0.2 M EDTA (pH 8.0) and/or heat to 65 °C.

Application

  • T7 RNA Polymerase can transcribe RNA from cloned DNA templates that are downstream from a T7 promoter. The synthesis can be performed with labeled NTPs to generate highly labeled RNA. Synthesized RNA can be used in many applications, including: RNA or DNA blotting techniques
  • In situ hybridization
  • RNase protection studies: Transcripts synthesized by the enzyme are used as precursor RNA for studies on RNA splicing and processing.
  • Synthesis of capped RNA in vitro with addition of m7GpppG or m7GpppA in excess over GTP or ATP during the transcription reaction. The generated antisense RNA can be introduced into cells to suppress the expression of the corresponding genes.
  • Microarray target synthesis

Packaging

1 kit containing 2 components

Unit Definition

One unit is the enzyme activity which incorporates 1 nmol CMP in acid-precipitable RNA products within 60 minutes at +37 °C.

Volume Activity: ≥20 U/μl

Preparation Note

Activator: The T7 RNA polymerases are strongly stimulated by BSA or spermidine.

Storage and Stability

Keep container tightly closed in a dry and well-ventilated place.

Other Notes

Test Buffer
40 mM Tris-HCl, pH 8.0 (+20°C), 6 mM MgCl2, 10 mM dithiothreitol, 2 mM spermidine, pH approximately 8.0 (+20°C).
Absence of Endonucleases
1. 1 μg lambda DNA is incubated with T7 RNA polymerase for 4 hours at +37°C in 25 μl test buffer. The number of enzyme units which show no degradation of lambda DNA is >100 U.
2. 1 μg Eco RI/Hind III fragments of lambda DNA is incubated with T7 RNA polymerase for 4 hours at +37°C in 25 μl test buffer. The number of enzyme units which show no alteration of the banding pattern is >100 U.
Absence of Nicking Activity
1 μg pBR322 DNA is incubated with T7 RNA polymerase for 4 hours at +37°C in 25 μl test buffer. The number of enzyme units which show no relaxing of supercoiled structure is >100 U.
Absence of RNases
4 μg MS2 RNA are incubated with T7 RNA polymerase for 4 hours at +37°C in 50 μl test buffer. The number of enzyme units which show no degradation of MS2 RNA is >100 U.
Performance in Transcription Assay
T7 RNA polymerase is function tested in the SP6/T7 Transcription Kit (Cat. No. 10 999 644 001). The incorporation rate in the standard assay with 0.5 μg pSPT19 neo DNA linearized with Eco RI and 50 mCi [alpha-32P] CTP, [400 Ci/mmol (15 TBq/mmol)] gives >50% of the input radioactivity in 20 minutes.
For life science research only. Not for use in diagnostic procedures.
Roche has 10x concentrated RNA Labeling Mixes that are specially designed for DIG- or biotin-labeling. These mixes work well with T7 RNA Polymerase.

Kit Components Only

Product No.
Description

  • T7 RNA Polymerase, in buffer, pH 7.9 ≥20 U/μl

  • Transcription Buffer 10x concentrated

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aditi Kulkarni et al.
Journal of genomics, 8, 30-36 (2020-03-20)
In the CRISPR-Cas systems, Cas13a is an RNA-guided RNA nuclease specifically targeting single strand RNA. We developed a Cas13a mediated CRISPR interference tool to target mRNA for gene silencing in mosquitoes. A Cas13a expressing plasmid was delivered to mosquitoes by
Kaia Achim et al.
Nature biotechnology, 33(5), 503-509 (2015-04-14)
Understanding cell type identity in a multicellular organism requires the integration of gene expression profiles from individual cells with their spatial location in a particular tissue. Current technologies allow whole-transcriptome sequencing of spatially identified cells but lack the throughput needed
Aditi Kulkarni et al.
Scientific reports, 12(1), 6005-6005 (2022-04-11)
Immune responses require delicate controls to maintain homeostasis while executing effective defense. Aryl hydrocarbon receptor (AhR) is a ligand-activated transcription factor. The Krüppel-like factor 10 (KLF10) is a C2H2 zinc-finger containing transcription factor. The functions of mosquito AhR and KLF10
Nicole Rosskothen-Kuhl et al.
PloS one, 9(3), e92624-e92624 (2014-03-22)
Brain development and learning is accompanied by morphological and molecular changes in neurons. The growth associated protein 43 (Gap43), indicator of neurite elongation and synapse formation, is highly expressed during early stages of development. Upon maturation of the brain, Gap43
Camilo Riquelme-Guzmán et al.
eLife, 11 (2022-10-12)
Early events during axolotl limb regeneration include an immune response and the formation of a wound epithelium. These events are linked to a clearance of damaged tissue prior to blastema formation and regeneration of the missing structures. Here, we report

Articles

T7 RNA Polymerase synthesizes RNA from DNA with high specificity, valuable for in vitro translation and anti-sense RNA.

T7 RNA Polymerase synthesizes RNA from DNA with high specificity, valuable for in vitro translation and anti-sense RNA.

T7 RNA Polymerase synthesizes RNA from DNA with high specificity, valuable for in vitro translation and anti-sense RNA.

T7 RNA Polymerase synthesizes RNA from DNA with high specificity, valuable for in vitro translation and anti-sense RNA.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service