Skip to Content
Merck
All Photos(1)

Key Documents

H1138

Sigma-Aldrich

Horse Serum

Donor Herd, USA origin, Heat inactivated, sterile-filtered, suitable for cell culture

Synonym(s):

HS, equine sera, equine serum, horse sera, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

biological source

horse serum

Quality Level

sterility

sterile-filtered

composition

Hemoglobin, ≤20 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL Endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Horse Serum has been used:
  • as a media supplement in the differentiation medium to culture the pheochromocytoma (PC12) cells
  • in immunocytochemistry and immunohistichemistry
  • to study the therapeutic effect of serum treatment on cardiomyocyte viability

Preparation Note

Collected from a controlled herd.
Prepared by heating at 56 °C for 30 minutes.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joerg Jores et al.
The Journal of infectious diseases, 219(10), 1559-1563 (2018-12-13)
Capsular polysaccharides have been confirmed to be an important virulence trait in many gram-positive and gram-negative bacteria. Similarly, they are proposed to be virulence traits in minimal Mycoplasma that cause disease in humans and animals. In the current study, goats
N Saito et al.
The Journal of infection, 46(1), 49-55 (2002-12-31)
Helicobacter pylori (H. pylori), present in a half of the world's population, is a very successful pathogen. The infection by this bacterium causes several gastric diseases in human. H. pylori is morphologically divided into two types; a spiral and a
Nathan J Marchant et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(22), 7447-7457 (2014-05-30)
In human alcoholics, abstinence is often self-imposed, despite alcohol availability, because of the negative consequences of excessive use. During abstinence, relapse is often triggered by exposure to contexts associated with alcohol use. We recently developed a rat model that captures
Julien Dufort-Gervais et al.
Scientific reports, 10(1), 6956-6956 (2020-04-26)
Synapse loss occurs early and correlates with cognitive decline in Alzheimer's disease (AD). Synaptotoxicity is driven, at least in part, by amyloid-beta oligomers (Aβo), but the exact synaptic components targeted by Aβo remain to be identified. We here tested the
Gianluca Fulgenzi et al.
Nature communications, 11(1), 1950-1950 (2020-04-25)
BDNF signaling in hypothalamic circuitries regulates mammalian food intake. However, whether BDNF exerts metabolic effects on peripheral organs is currently unknown. Here, we show that the BDNF receptor TrkB.T1 is expressed by pancreatic β-cells where it regulates insulin release. Mice

Articles

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service