Skip to Content
Merck
All Photos(1)

Key Documents

11426338910

Roche

Anti-Fluorescein-AP, Fab fragments

from sheep

Synonym(s):

antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

sheep

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

solution

packaging

pkg of 150 U (200 μl)

manufacturer/tradename

Roche

isotype

IgG

storage temp.

2-8°C

General description

Fluorescein is synthesized from m-dialkylaminophenols or resorcinols condensed with phthalic anhydrides under harsh conditions. Fab fragments bind to G proteins.
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional activator after forming a protein complex with other proteins. This protein acts as a nucleocytoplasmic shuttle protein and is important for neural crest and peripheral nervous system development. Mutations in this gene are associated with Waardenburg-Shah and Waardenburg-Hirschsprung disease. (provided by Ref Seq)

Specificity

The polyclonal antibody reacts with free and bound fluorescein.

Application

Use Anti-Fluorescein-AP, Fab fragments for the detection of fluorescein-labeled compounds using:
  • Dot blot
  • ELISA
  • Immunohistocytochemistry
  • In situ hybridization
  • Southern blot
  • Western blot
  • Electrochemical biosensing technique for the detection of 16S rRNA

Biochem/physiol Actions

Anti-Fluorescein-AP, Fab fragments has been used in oligonucleotide capture, denaturation and detection (CDD).

Features and Benefits

Contents
Solution, stabilized

Preparation Note

  • Working concentration: The following concentrations depend on application and substrate and should be taken as a guideline. Dot blot: 150mU/ml
  • ELISA: 150 to 300mU/ml
  • Immunohistocytochemistry: 250 to 500mU/ml
  • In situ hybridization: 1.5 to 7.5U/ml
  • Southern blot: 150mU/ml
  • Western blot: 250 to 500 mU/ml

Working solution: 100mM Tris-HCl, 150mM NaCl, pH 7.5. If necessary 1% Blocking reagent (w/v), dry milk powder, 1 to 5% heat inactivated fetal calf serum (v/v) or sheep normal serum can be used for reduction of unspecific binding.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana L Miranda-Angulo et al.
The Journal of comparative neurology, 522(4), 876-899 (2013-08-14)
The wall of the ventral third ventricle is composed of two distinct cell populations: tanycytes and ependymal cells. Tanycytes regulate many aspects of hypothalamic physiology, but little is known about the transcriptional network that regulates their development and function. We
Fluorescent indicators for cytosolic calcium based on rhodamine and fluorescein chromophores
Minta A, et al.
The Journal of Biological Chemistry, 264(14), 8171-8178 (1989)
Simplified Paper Format for Detecting HIV Drug Resistance in Clinical Specimens by Oligonucleotide Ligation
Panpradist N, et al.
PLoS ONE (2016)
Mandy L Y Sin et al.
Journal of microelectromechanical systems : a joint IEEE and ASME publication on microstructures, microactuators, microsensors, and microsystems, 22(5), 1126-1132 (2014-05-27)
Transforming microfluidics-based biosensing systems from laboratory research into clinical reality remains an elusive goal despite decades of intensive research. A fundamental obstacle for the development of fully automated microfluidic diagnostic systems is the lack of an effective strategy for combining
Megan Addison et al.
Developmental cell, 45(5), 606-620 (2018-05-08)
The patterning of tissues to form subdivisions with distinct and homogeneous regional identity is potentially disrupted by cell intermingling. Transplantation studies suggest that homogeneous segmental identity in the hindbrain is maintained by identity switching of cells that intermingle into another segment. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service