Skip to Content
Merck
All Photos(4)

Key Documents

P6280

Sigma-Aldrich

Pyridoxine hydrochloride

BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture

Synonym(s):

Adermine hydrochloride, PN HCl, Pyridoxol hydrochloride, Vitamin B6 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO3 · HCl
CAS Number:
Molecular Weight:
205.64
Beilstein:
3632435
EC Number:
MDL number:
UNSPSC Code:
12161900
eCl@ss:
34058008
PubChem Substance ID:
NACRES:
NA.21

product line

BioReagent

Quality Level

Assay

≥98% (HPLC)

form

powder

mol wt

Mw 205.64 g/mol

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable
cell culture | plant: suitable

color

white to off-white

mp

214-215 °C (lit.)

solubility

water: soluble 50 mg/mL, clear, colorless to faintly yellow

application(s)

agriculture

SMILES string

Cl[H].Cc1ncc(CO)c(CO)c1O

InChI

1S/C8H11NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,10-12H,3-4H2,1H3;1H

InChI key

ZUFQODAHGAHPFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyridoxine is a water-soluble vitamin that exists in six forms called, pyridoxal, pyridoxine, pyridoxamine, pyridoxamine 5′-phosphate, pyridoxine 5′-phosphate, and pyridoxal 5′-phosphate. 

Application

Pyridoxine hydrochloride has been used:
  • as a vitamin component in a chemically defined diet with balanced metal content for metal depletion studies
  • as a component of chemically defined medium (CDM) to culture Staphylococcus aureus WSPPA
  • as a vitamin to study the impact of protein expression and post-translational modifications specifically charge heterogeneity

Biochem/physiol Actions

Pyridoxine functions as an essential coenzyme for amino acid, carbohydrate, and lipid metabolism. Pyridoxine deficiency is associated with peripheral neuropathy, microcytic anemia, and electroencephalographic abnormalities. It is implicated in a wide range of biochemical reactions that affect cellular processes. Pyridoxine serves as an alternative to pyridoxal which can degrade in media.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carmela Vigorito et al.
International journal of molecular sciences, 20(9) (2019-05-11)
(1) The beneficial effects of hydrogen sulfide (H2S) on the cardiovascular and nervous system have recently been re-evaluated. It has been shown that lanthionine, a side product of H2S biosynthesis, previously used as a marker for H2S production, is dramatically
Wanliang Shi
Antibiotics (Basel, Switzerland), 10(8) (2021-08-28)
Susceptibility testing of tuberculosis (TB) drugs on Mycobacterium tuberculosis is essential for the rapid detection of strains resistant to the drugs, providing the patient with effective treatment, and preventing the spread of drug-resistant TB strains. Pyrazinamide (PZA) is one of
Imène Mensi et al.
Bio-protocol, 7(2), e2111-e2111 (2017-01-20)
Sugarcane (interspecific hybrids of Saccharum species) is an economically important crop that provides 70% of raw table sugar production worldwide and contributes, in some countries, to bioethanol and electricity production. Leaf scald, caused by the bacterial plant pathogen Xanthomonas albilineans
Manfred Eggersdorfer et al.
Angewandte Chemie (International ed. in English), 51(52), 12960-12990 (2012-12-05)
The discovery of vitamins as essential factors in the diet was a scientific breakthrough that changed the world. Diseases such as scurvy, rickets, beriberi, and pellagra were recognized to be curable with an adequate diet. These diseases had been prevalent
Vijay P Singh et al.
The Journal of organic chemistry, 78(4), 1478-1487 (2013-01-16)
Vitamin B6 is involved in a variety of enzymatic transformations. Some recent findings also indicate an antioxidant role of the vitamin in biological systems. We set out to turn pyridoxine (1a) into a catalytic chain-breaking and hydroperoxide-decomposing antioxidant by replacing

Articles

Vitamin B6's impact on serum-free cell culture systems is explored for protein manufacturing.

Vitamin B6's impact on serum-free cell culture systems is explored for protein manufacturing.

Vitamin B6's impact on serum-free cell culture systems is explored for protein manufacturing.

Vitamin B6's impact on serum-free cell culture systems is explored for protein manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service