Merck
  • Activity of Pyrazinamide against Mycobacterium tuberculosis at Neutral pH in PZA-S1 Minimal Medium.
Activity of Pyrazinamide against Mycobacterium tuberculosis at Neutral pH in PZA-S1 Minimal Medium.
Antibiotics (Basel, Switzerland) (2021-08-28)
Wanliang Shi
ABSTRACT
Susceptibility testing of tuberculosis (TB) drugs on Mycobacterium tuberculosis is essential for the rapid detection of strains resistant to the drugs, providing the patient with effective treatment, and preventing the spread of drug-resistant TB strains. Pyrazinamide (PZA) is one of the first-line agents used for the treatment of TB. However, current phenotypic PZA susceptibility testing is unreliable due to its performance in acidic pH conditions. The aims of this study were to develop minimal media to determine the activity of PZA at a neutral pH at 37 °C to avoid problems caused by an acidic pH, which is currently used in PZA susceptibility tests, and to identify PZA-resistant M. tuberculosis in media with reproducibility and accuracy. Different minimal media were used to determine the activity of PZA using the broth microdilution method with M. tuberculosis H37Ra as the reference strain. The PZA-S1 minimal medium was proposed as the most suitable medium. PZA inhibited the growth of M. tuberculosis in PZA-S1 at a neutral pH of 6.8, which is the optimal pH for M. tuberculosis growth. Moreover, PZA showed activity at a neutral pH on a PZA-S1 agar plate when using the disk diffusion method. PZA-resistant M. tuberculosis could be identified at a neutral pH in PZA-S1 minimal medium. This study establishes valuable information regarding the testing of PZA's susceptibility in relation to M. tuberculosis at a neutral pH of 6.8 with reliability and accuracy in clinical settings.
Sigma-Aldrich
Pyridoxine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Zinc sulfate heptahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Ammonium iron(III) citrate, reagent grade, powder
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%