Merck
One hundred years of vitamins-a success story of the natural sciences.
Angewandte Chemie (International ed. in English) (2012-12-05)
Manfred Eggersdorfer, Dietmar Laudert, Ulla Létinois, Tom McClymont, Jonathan Medlock, Thomas Netscher, Werner Bonrath
ABSTRACT
The discovery of vitamins as essential factors in the diet was a scientific breakthrough that changed the world. Diseases such as scurvy, rickets, beriberi, and pellagra were recognized to be curable with an adequate diet. These diseases had been prevalent for thousands of years and had a dramatic impact on societies as well as on economic development. This Review highlights the key achievements in the development of industrial processes for the manufacture of eight of the 13 vitamins.
Product Description
Supelco
Pyridoxine Hydrochloride (B6), analytical standard
Sigma-Aldrich
Pyridoxine hydrochloride, meets USP testing specifications
Sigma-Aldrich
Pyridoxine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Pyridoxine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Pyridoxine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyridoxine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Pyridoxine, ≥98%