Skip to Content
Merck
All Photos(1)

Key Documents

T8154

Sigma-Aldrich

Trypan Blue solution

0.4%, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Blue cell viability dye

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H24N6Na4O14S4
CAS Number:
Molecular Weight:
960.81
Beilstein:
4360496
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

sterility

sterile-filtered

Quality Level

form

liquid

storage condition

dry at room temperature

concentration

0.4%

technique(s)

cell culture | mammalian: suitable
tissue processing: suitable

application(s)

cell analysis

shipped in

ambient

SMILES string

[Na+].[Na+].[Na+].[Na+].Cc1cc(ccc1N=Nc2c(O)c3c(N)cc(cc3cc2S([O-])(=O)=O)S([O-])(=O)=O)-c4ccc(N=Nc5c(O)c6c(N)cc(cc6cc5S([O-])(=O)=O)S([O-])(=O)=O)c(C)c4

InChI

1S/C34H28N6O14S4.4Na/c1-15-7-17(3-5-25(15)37-39-31-27(57(49,50)51)11-19-9-21(55(43,44)45)13-23(35)29(19)33(31)41)18-4-6-26(16(2)8-18)38-40-32-28(58(52,53)54)12-20-10-22(56(46,47)48)14-24(36)30(20)34(32)42;;;;/h3-14,41-42H,35-36H2,1-2H3,(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;/q;4*+1/p-4

InChI key

GLNADSQYFUSGOU-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Trypan Blue (TB), an anionic hydrophilic azo dye, crosses only the cell membranes of dead cells, thereby staining dead tissues/cells blue. It is used in quantitative microscopy for a dye exclusion staining technique to differentiate between live and dead mammalian cells.
Trypan blue is a dye used to distinguish between live and dead cells. It is a vital stain that is not absorbed by healthy viable cells, but stains cells with a damaged cell membrane. This way, only dead cells can be counted. The method is sometimes referred to as the dye exclusion method.

Application

Trypan Blue solution has been used:
  • in cell viability assay to count viable and dead cells
  • in rescue assay to count viable cells
  • in trypan blue exclusion counting method to determine proliferation curve
  • as the saline control and in the preparation of lipopolysaccharide (LPS) solution to confirm the success of LPS infusion
  • in trypan blue dye exclusion test to determine gastric epithelial cells (GEC) viability
  • to count GEC cells using a hemocytometer

Preparation Note

Prepared in 0.81% sodium chloride and 0.06% potassium phosphate, dibasic.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single cell isolation process with laser induced forward transfer
Yu Deng
Journal of Biological Engineering, 11(2) (2017)
The sensitivity of 5-formylcytosine to doxorubicin regardless of DNA damage
Selcen
Turkiye Entomoloji Dergisi (Turkish Journal of Entomology), 40 (2016)
Vital dyes in vitreoretinal surgery: chromovitrectomy
Meyer CH
Developments in Ophthalmology, 42 (2008)
Roya Khosravi-Far, Zahra Zakeri
Programmed Cell Death (2008)
Ulrich Putz et al.
Science signaling, 5(243), ra70-ra70 (2012-09-27)
Exosomes are microvesicles of endosomal origin that are secreted, and their contents (proteins, lipids, DNA, or microRNAs) can alter the physiological states of recipient cells. We demonstrated that phosphatase and tensin homolog deleted on chromosome 10 (PTEN), a tumor suppressor

Articles

Cell counting protocol using a hemocytometer for monitoring cell viability. Discover other cell counting tools such as the Scepter™ 3.0 Cell Counter.

Cell counting protocol using a hemocytometer for monitoring cell viability. Discover other cell counting tools such as the Scepter™ 3.0 Cell Counter.

Cell counting protocol using a hemocytometer for monitoring cell viability. Discover other cell counting tools such as the Scepter™ 3.0 Cell Counter.

Cell counting protocol using a hemocytometer for monitoring cell viability. Discover other cell counting tools such as the Scepter™ 3.0 Cell Counter.

Protocols

Cell culture protocol for passaging and splitting adherent cell lines using trypsin EDTA. Free ECACC handbook download.

Cell culture protocol for passaging and splitting adherent cell lines using trypsin EDTA. Free ECACC handbook download.

Cell culture protocol for passaging and splitting adherent cell lines using trypsin EDTA. Free ECACC handbook download.

Cell culture protocol for passaging and splitting adherent cell lines using trypsin EDTA. Free ECACC handbook download.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service