Skip to Content
Merck
All Photos(3)

Key Documents

A2934

Sigma-Aldrich

Bovine Serum Albumin

lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27

biological source

bovine

Quality Level

Assay

≥98% (agarose gel electrophoresis)

form

lyophilized powder

mol wt

~66 kDa

purified by

heat shock fractionation

packaging

poly bottle of

origin

USA origin

technique(s)

ELISA: suitable

impurities

essentially globulin free
≤1 ng/mg endotoxin

pH

5.2

solubility

water: soluble (40 mg/ml)

UniProt accession no.

foreign activity

BT Virus, none detected
VSV Virus, none detected

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Bovine Serum Albumin (BSA) is a globular non-glycosylated protein that encompasses structurally distinct domains (I, II, and III) with two sub-domains, A and B within each domain. Structurally, BSA is an α-helical protein with 17 cysteine residues.

Application

Bovine Serum Albumin has been used:
  • as a component of the minimal liquid medium to culture Porphyromonas gingivalis strains
  • as a standard protein to quantify microsomes homogenate using Bradford method
  • as a blocking agent in enzyme-linked immunosorbent assay (ELISA) of fecal lactoferrin

Biochem/physiol Actions

Bovine Serum Albumin (BSA) is widely used in drugs and antibody delivery. It also finds applications in nanoparticle preparations and is useful in targeting tumor tissues. BSA is non-immunogenic and non-toxic. It also shows binding with metal ions.
Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Features and Benefits

  • Low endotoxin
  • Essentially globulin-free

Preparation Note

Prepared using heat shock fractionation
Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nancy Machorro-Rojas et al.
Molecular medicine reports, 20(3), 2083-2090 (2019-07-02)
The intestinal epithelium is a monolayer of cells arranged side‑by‑side and connected by tight junction (TJ) proteins expressed at the apical extreme of the paracellular membrane. This layer prevents stress‑induced inflammatory responses, thus helping to maintain gut barrier function and
Botros B Shenoda et al.
Current protocols in pharmacology, 79, 9-9 (2017-12-21)
Pharmacogenomic approaches used to investigate how genes affect drug responses are critical for designing personalized therapies aimed at maximizing efficacy and minimizing adverse effects. Drug efficacy is often dependent on the sequence and expression levels of drug target genes or
Tamara Topală et al.
Clujul medical (1957), 87(4), 215-219 (2014-01-01)
The continuous search for new molecules with therapeutic abilities has led to the synthesis and characterization of a large number of metal complexes, proven to exhibit potential as pharmacological agents through their antibacterial, antiviral, antifungal and antineoplastic properties. As serum
Balázs Rada et al.
Journal of immunology (Baltimore, Md. : 1950), 181(7), 4883-4893 (2008-09-20)
The dual oxidase-thiocyanate-lactoperoxidase (Duox/SCN(-)/LPO) system generates the microbicidal oxidant hypothiocyanite in the airway surface liquid by using LPO, thiocyanate, and Duox-derived hydrogen peroxide released from the apical surface of the airway epithelium. This system is effective against several microorganisms that
Marcos V A S Navarro et al.
PLoS biology, 9(2), e1000588-e1000588 (2011-02-10)
The bacterial second messenger bis-(3'-5') cyclic dimeric guanosine monophosphate (c-di-GMP) has emerged as a central regulator for biofilm formation. Increased cellular c-di-GMP levels lead to stable cell attachment, which in Pseudomonas fluorescens requires the transmembrane receptor LapD. LapD exhibits a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service