Skip to Content
Merck
All Photos(2)

Key Documents

A0336

Sigma-Aldrich

Bovine Serum Albumin solution

30% in saline, IgG free, aseptically filled

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27

biological source

bovine

Quality Level

sterility

aseptically filled

Assay

28-32% protein basis (biuret)

form

solution

purified by

heat shock fractionation

does not contain

stabilizer or preservative

origin

USA origin

concentration

28-32% (biuret)
30% in saline

technique(s)

cell culture | mammalian: suitable

impurities

≤25 ng/mg protein IgG

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Albumin is the most abundant plasma protein in humans. High levels of albumin are associated with dehydration. Albumin turnover is seen in infants with iron deficiency anemia. Serum albumin is a reliable prognostic indicator for liver disease. Oxidative damage of albumin is associated with advanced liver disease.

Application

Bovine Serum Albumin solution has been used as a component:
  • of Eagle′s minimal essential medium (MEM) used as the liquid maintenance medium (MM) for virus infection
  • of blocking solution in stimulated and automated immunostaining
  • of Temin′s modified eagles medium to prepare the fresh overlay medium
  • in binding medium to perform binding assays

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Implementation of high-content assay for inhibitors of mitogen-activated protein kinase phosphatases
Vogt A and Lazo JS
Methods, 42(3), 268-277 (2007)
Matheswaran Kandasamy et al.
eLife, 12 (2023-01-11)
Virus-based tumour vaccines offer many advantages compared to other antigen-delivering systems. They generate concerted innate and adaptive immune response, and robust CD8+ T cell responses. We engineered a non-replicating pseudotyped influenza virus (S-FLU) to deliver the well-known cancer testis antigen
Infected erythrocytes expressing DC13 PfEMP1 differ from recombinant proteins in EPCR-binding function
Azasi Y, et al.
Proceedings of the National Academy of Sciences of the USA, 115(5), 1063-1068 (2018)
The role of fresh cow's milk in iron deficiency: I. Albumin turnover in infants with iron deficiency anemia
Woodruff C W and Clark J L
American Journal of Diseases of Children, 124, 18-23 (1972)
New low-viscosity overlay medium for viral plaque assays
Matrosovich M, et al.
Virology Journal, 3(1), 63-63 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service