Skip to Content
Merck
All Photos(5)

Key Documents

A3059

Sigma-Aldrich

Bovine Serum Albumin

heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27

biological source

bovine

Quality Level

Assay

≥98%

form

lyophilized powder

mol wt

~66 kDa

purified by

heat shock fractionation

packaging

poly bottle of

origin

USA origin

technique(s)

ELISA: suitable
western blot: suitable

impurities

essentially globulin free
protease, none detected

loss

≤5%

pH

7

solubility

water: soluble (40 mg/ml)

UniProt accession no.

foreign activity

BT Virus, none detected
VSV Virus, none detected

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Bovine serum albumin (BSA) has an important role in the regulation of blood colloidal osmotic pressure. BSA is the major protein in bovine serum, and is composed of 583 amino acids with a calculated molecular weight of 66,430 Daltons. BSA is used in many different applications and fields, such as:
  • Protein quantitation (as a standard)
  • Immunochemistry (as a blocking agent)
  • Cell culture (as a nutrient)

This particular BSA is specifically evaluated and tested to have very low protease content / for protease-free status. Protease-free BSA may be used in applications like:
  • Enzyme assays
  • Protein-based assays
  • Protease-sensitive techniques that include:
a) Enzyme immunoassay (EIA)
b) Nucleic acid hybridization
c) Radioimmunoassay (RIA)

Application

Bovine Serum Albumin has been used in luminescence assays.

Biochem/physiol Actions

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Features and Benefits

  • Heat shock fractionated
  • Protease-free
  • Essentially globulin-free

Preparation Note

Prepared using heat shock fractionation
Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dual luminescence-based reporter gene assay for luciferase and beta-galactosidase
Martin C S, et al.
Biotechniques, 520-524 (1996)
Priyank A Shenoy et al.
Frontiers in pharmacology, 9, 495-495 (2018-06-06)
In the majority of patients with breast cancer in the advanced stages, skeletal metastases are common, which may cause excruciating pain. Currently available drug treatments for relief of breast cancer-induced bone pain (BCIBP) include non-steroidal anti-inflammatory drugs and strong opioid
Edith L Pfister et al.
Human gene therapy, 29(6), 663-673 (2017-12-07)
Huntington's disease (HD) is a fatal neurodegenerative disease caused by a genetic expansion of the CAG repeat region in the huntingtin (HTT) gene. Studies in HD mouse models have shown that artificial miRNAs can reduce mutant HTT, but evidence for
Leiting Pan et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(18), 1900865-1900865 (2019-09-29)
The dynamic response of the cell to osmotic changes is critical to its physiology and is widely exploited for cell manipulation. Here, using three-dimensional stochastic optical reconstruction microscopy (3D-STORM), a super-resolution technique, the hypotonic stress-induced ultrastructural changes of the cytoskeleton
Amanda Phuong Tran et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 38(23), 5399-5414 (2018-05-16)
Severed axon tips reform growth cones following spinal cord injury that fail to regenerate, in part, because they become embedded within an inhibitory extracellular matrix. Chondroitin sulfate proteoglycans (CSPGs) are the major axon inhibitory matrix component that is increased within

Related Content

Learn about bovine serum albumin (BSA) structure, molecular weight, function, solubility, how BSA is made, purification methods, applications and purchasing.

Learn about bovine serum albumin (BSA) structure, molecular weight, function, solubility, how BSA is made, purification methods, applications and purchasing.

Learn about bovine serum albumin (BSA) structure, molecular weight, function, solubility, how BSA is made, purification methods, applications and purchasing.

Learn about bovine serum albumin (BSA) structure, molecular weight, function, solubility, how BSA is made, purification methods, applications and purchasing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service