Skip to Content
Merck
All Photos(5)

Key Documents

A4161

Sigma-Aldrich

Bovine Serum Albumin

lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27

biological source

bovine

Quality Level

product line

BioReagent

Assay

≥99%

form

lyophilized powder

mol wt

~66 kDa

purified by

cold ethanol and heatshock fractionation

packaging

poly bottle of

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

essentially globulin free

pH

7

solubility

water: soluble (50 mg/ml)

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Features and Benefits

  • Tested for use in cell culture
  • Essentially globulin free

Preparation Note

Purified by a combination of heat shock and ethanol fractionation
Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guido Bolognesi et al.
Nature communications, 9(1), 1882-1882 (2018-05-16)
Constructing higher-order vesicle assemblies has discipline-spanning potential from responsive soft-matter materials to artificial cell networks in synthetic biology. This potential is ultimately derived from the ability to compartmentalise and order chemical species in space. To unlock such applications, spatial organisation
Mark A Petersen et al.
Neuron, 96(5), 1003-1012 (2017-11-07)
Blood-brain barrier (BBB) disruption alters the composition of the brain microenvironment by allowing blood proteins into the CNS. However, whether blood-derived molecules serve as extrinsic inhibitors of remyelination is unknown. Here we show that the coagulation factor fibrinogen activates the
Melisa Herrero et al.
Neuromolecular medicine, 21(3), 303-313 (2019-05-28)
Vanishing white matter (VWM) disease (OMIM#306896) is an autosomal recessive neurodegenerative leukodystrophy caused by hypomorphic mutations in any of the five genes encoding the subunits of eukaryotic translation initiation factor 2B (eIF2B). The disease is manifested by loss of cerebral
Aseel El Hajj et al.
PloS one, 14(6), e0218812-e0218812 (2019-06-25)
The regulation of cholesterol, an essential brain lipid, ensures proper neuronal development and function, as demonstrated by links between perturbations of cholesterol metabolism and neurodegenerative diseases, including Alzheimer's disease. The central nervous system (CNS) acquires cholesterol via de novo synthesis
Silvana Konermann et al.
Cell, 173(3), 665-676 (2018-03-20)
Class 2 CRISPR-Cas systems endow microbes with diverse mechanisms for adaptive immunity. Here, we analyzed prokaryotic genome and metagenome sequences to identify an uncharacterized family of RNA-guided, RNA-targeting CRISPR systems that we classify as type VI-D. Biochemical characterization and protein

Articles

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Related Content

Learn about bovine serum albumin (BSA) structure, molecular weight, function, solubility, how BSA is made, purification methods, applications and purchasing.

Learn about bovine serum albumin (BSA) structure, molecular weight, function, solubility, how BSA is made, purification methods, applications and purchasing.

Learn about bovine serum albumin (BSA) structure, molecular weight, function, solubility, how BSA is made, purification methods, applications and purchasing.

Learn about bovine serum albumin (BSA) structure, molecular weight, function, solubility, how BSA is made, purification methods, applications and purchasing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service