Recommended Products
Product Name
Phosphate buffered saline, tablet
form
tablet
solubility
H2O: 1 tablet/200 mL, clear, colorless
application(s)
diagnostic assay manufacturing
Looking for similar products? Visit Product Comparison Guide
Reconstitution
Application procedure:
Dissolve 1 tablet in 200 mL water to obtain a 137 mM NaCl, 2.7 mM KCl and 10 mM phosphate buffer solution (pH 7.4 at 25 °C)
Dissolve 1 tablet in 200 mL water to obtain a 137 mM NaCl, 2.7 mM KCl and 10 mM phosphate buffer solution (pH 7.4 at 25 °C)
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Leila Sabouri et al.
ACS biomaterials science & engineering, 6(11), 6285-6298 (2021-01-16)
The human amniotic membrane (HAM) has been viewed as a potential regenerative material for a wide variety of injured tissues because of its collagen-rich content. High degradability of HAM limits its wide practical application in bone tissue engineering. In this
Tzu-Jing Yang et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(3), 1438-1446 (2020-01-05)
Feline infectious peritonitis virus (FIPV) is an alphacoronavirus that causes a nearly 100% mortality rate without effective treatment. Here we report a 3.3-Å cryoelectron microscopy (cryo-EM) structure of the serotype I FIPV spike (S) protein, which is responsible for host
Sarit Anava et al.
Biophysical journal, 96(4), 1661-1670 (2009-02-17)
A bewildering series of dynamical processes take part in the development of the nervous system. Neuron branching dynamics, the continuous formation and elimination of neural interconnections, are instrumental in constructing distinct neuronal networks, which are the functional building blocks of
Michelina Stoddard et al.
Scientific reports, 9(1), 28-28 (2019-01-12)
Studying early immune responses to organ damage in situ requires animal models amenable to intravital imaging. Here, we used transparent zebrafish larvae, a powerful animal model for innate immunity, to measure leukocyte recruitment to damaged livers. Bath application of metronidazole
Yong N Li et al.
The Journal of comparative neurology, 516(5), 442-453 (2009-08-06)
Horizontal cells (HCs) are involved in establishing the center-surround receptive field organization of photoreceptor and bipolar cells. In many species, HCs respond differentially to colors and may play a role in color vision. An earlier study from our laboratory suggested
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service