Skip to Content
Merck
All Photos(1)

Key Documents

P3688

Sigma-Aldrich

Phosphate buffered saline

powder, pH 7.4, contains BSA

Synonym(s):

PBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41105319
eCl@ss:
32129211
NACRES:
NA.25

product name

Phosphate buffered saline, pH 7.4, contains BSA, powder

form

powder

Quality Level

contains

BSA

pH

7.4

solubility

water: soluble

application(s)

diagnostic assay manufacturing

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Phosphate buffered saline has been used for the dilution of lyotrophic liquid crystals (LLC) containing solution for the release studies. It has also been used as a blocking solution in enzyme linked immunosorbent assay (ELISA).

Packaging

Foil pouches

Reconstitution

Contents of one pouch, when dissolved in one liter of distilled or deionized water, will yield 0.01 M phosphate buffered saline (NaCl, 0.138 M; KCl, 0.0027 M); bovine serum albumin - 1% w/v, pH 7.4 at 25 °C.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determining in-field dispersal of the redbanded stink bug (Hemiptera: Pentatomidae) in soybean fields using a protein based mark-capture method
Bastola A and Davis JA
Crop Protection, 112, 24-32 (2018)
Structural characterization of lyotropic liquid crystals containing a dendrimer for solubilization and release of gallic acid
Bitan-Cherbakovsky L, et al.
Colloids and Surfaces. B, Biointerfaces, 112, 87-95 (2013)
Discovery of antivirulence agents against methicillin-resistant Staphylococcus aureus
Khodaverdian V, et al.
Antimicrobial agents and chemotherapy, 57(8), 3645-3652 (2013)
Yong N Li et al.
The Journal of comparative neurology, 516(5), 442-453 (2009-08-06)
Horizontal cells (HCs) are involved in establishing the center-surround receptive field organization of photoreceptor and bipolar cells. In many species, HCs respond differentially to colors and may play a role in color vision. An earlier study from our laboratory suggested
S Vaddiraju et al.
Biosensors & bioelectronics, 24(6), 1557-1562 (2008-10-01)
The performance of an implantable glucose sensor is strongly dependent on the ability of their outer membrane to govern the diffusion of the various participating species. In this contribution, using a series of layer-by-layer (LBL) assembled outer membranes, the role

Protocols

Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.

Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.

Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.

Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.

Related Content

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service