P5119
Phosphate buffered saline
liquid, pH 7.2 (25 °C)
Synonym(s):
PBS
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Product Name
Phosphate buffered saline, pH 7.2 (25 °C)
sterility
autoclaved
Quality Level
form
liquid
pH
7.2 (25 °C)
application(s)
diagnostic assay manufacturing
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Phosphate buffered saline (PBS) is an isotonic buffer solution. This water-based salt solution is made up of disodium hydrogen phosphate, sodium chloride and, in some preparations, potassium chloride and potassium dihydrogen phosphate. PBS is generally used in biological research.
Application
It has also been used for the dilution of capture antibody during α-hemolysin (Hla) enzyme-linked immunosorbent assay (ELISA).
Phosphate buffered saline has been used in bis-benzimide accumulation assay.
Components
Sodium chloride, 145 mM (0.85%) in phosphate buffer, 150 mM
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Experimental? review? of? cobalt? induced cardiomyopathy
Zadnipryany I, et al.
Russian Open Medical Journal, 6(1), 103-103 (2017)
ELISA for Alpha-hemolysin (Hla) in Methicilin-resistant Staphylococcus aureus (MRSA)
Khodaverdian VY and Shoham M
Antimicrobial agents and chemotherapy, 6 (2013)
Measuring the activity of active efflux in Gram-negative bacteria
Antibiotic resistance protocols, 173-180 (2010)
Daniela Michlmayr et al.
Cell reports, 31(4), 107569-107569 (2020-04-30)
Zika virus (ZIKV) is an emerging, mosquito-borne flavivirus responsible for recent epidemics across the Americas, and it is closely related to dengue virus (DENV). Here, we study samples from 46 DENV-naive and 43 DENV-immune patients with RT-PCR-confirmed ZIKV infection at
Yong N Li et al.
The Journal of comparative neurology, 516(5), 442-453 (2009-08-06)
Horizontal cells (HCs) are involved in establishing the center-surround receptive field organization of photoreceptor and bipolar cells. In many species, HCs respond differentially to colors and may play a role in color vision. An earlier study from our laboratory suggested
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service