Skip to Content
Merck
All Photos(1)

Key Documents

P7059

Sigma-Aldrich

Phosphate Buffered Saline

10X, liquid, pH 7.2-7.6 (1x solution), for molecular biology

Synonym(s):

Phosphate buffered saline, PBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
eCl@ss:
32129211
NACRES:
NA.25

product name

Phosphate Buffered Saline, 10× PBS for Western blots and IP

grade

for molecular biology

Quality Level

sterility

0.2 μm filtered

form

liquid

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

pH

7.2-7.6(1x solution)

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Phosphate Buffered Saline (PBS) is routinely used as a wash buffer in Western blot and immunoprecipitation (IP) procedures, as well as during sample preparation of cell lysates for use in general molecular biology applications.

Application

Suitable for use in wash buffers for Western blots and IP, as well as cell lysate sample preparation.

Components

10x PBS contains:
  • 30mM sodium phosphate
  • 1.5 M sodium chloride
  • 10.5 mM potassium phosphate

Preparation Note

Dilute 10x PBS to a 1x working solution using molecular biology grade water.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sahar M Greish et al.
International journal of physiology, pathophysiology and pharmacology, 12(5), 134-146 (2020-11-24)
Hypertension can be induced by inhibiting nitric oxide synthesis with L-NAME, which also has a role in oxidative stress. Curcumin has strong antioxidant property. Our aim was to examine the possible preventive role of curcumin on renal dysfunction secondary to
Jennifer L S Sporty et al.
Analytical chemistry, 82(15), 6593-6600 (2010-07-14)
A novel method for extracting butyrylcholinesterase (BuChE) from serum as a means of identifying and measuring nerve agent adducts to human BuChE is presented here. Antibutyrylcholinesterase monoclonal antibodies were conjugated to protein-G ferromagnetic particles and mixed with 500 microL serum
Johan E Karlsson et al.
Drug metabolism and disposition: the biological fate of chemicals, 38(4), 705-714 (2010-01-15)
Membrane-bound transporter proteins play an important role in the efflux of drugs from cells and can significantly influence the pharmacokinetics of drug molecules. This study describes the production of large amounts of high-activity transporter membrane vesicles from human embryonic kidney
Shahani Noor et al.
Brain, behavior, and immunity, 61, 80-95 (2016-12-25)
A growing body of evidence indicates that prenatal alcohol exposure (PAE) may predispose individuals to secondary medical disabilities later in life. Animal models of PAE reveal neuroimmune sequelae such as elevated brain astrocyte and microglial activation with corresponding region-specific changes
Katie Burbridge et al.
Development (Cambridge, England), 148(8) (2021-04-30)
Body tissues are frequently exposed to stress, from toxic byproducts generated during cellular metabolism through to infection or wounding. Although it is well-established that tissues respond to exogenous injury by rapidly upregulating cytoprotective machinery, how energetically demanding tissues - vulnerable

Articles

Learn how water impurities can affect pH. Water quality defined as pure water or Type 2 water is recommended to prepare buffer solutions or measure pH.

Learn how water impurities can affect pH. Water quality defined as pure water or Type 2 water is recommended to prepare buffer solutions or measure pH.

Learn how water impurities can affect pH. Water quality defined as pure water or Type 2 water is recommended to prepare buffer solutions or measure pH.

Learn how water impurities can affect pH. Water quality defined as pure water or Type 2 water is recommended to prepare buffer solutions or measure pH.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service