Skip to Content
Merck
All Photos(1)

Key Documents

47739

Sigma-Aldrich

D-(−)-Fructose

BioUltra, ≥99.0% (HPLC)

Synonym(s):

D-Levulose, Fruit sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1239004
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Quality Level

Assay

≥99.0% (HPLC)

form

solid

optical activity

[α]20/D −92±2°, 1 hr, c = 10% in H2O

impurities

insoluble matter, passes filter test

ign. residue

≤0.1% (as SO4)

loss

≤0.1% loss on drying, 20 °C (HV)

color

colorless

pH

5.0-7.0 (25 °C, 0.1 M in H2O)

mp

119-122 °C (dec.) (lit.)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.1 M in H2O

UV absorption

λ: 260 nm Amax: 0.04
λ: 280 nm Amax: 0.04

storage temp.

room temp

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m1/s1

InChI key

BJHIKXHVCXFQLS-UYFOZJQFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-Fructose is an important monosaccharide used in a wide range of applications. D-fructose is used in carbohydrate research, in organic and bioorganic synthesis; as a model compound; as a reference material, as an enzyme substrate and as a nutrient.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Barclay et al.
Carbohydrate research, 347(1), 136-141 (2011-12-02)
D-Fructose was analysed by NMR spectroscopy and previously unidentified (1)H NMR resonances were assigned to the keto and α-pyranose tautomers. The full assignment of shifts for the various fructose tautomers enabled the use of (1)H NMR spectroscopy in studies of
Young Ki Kim et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-19)
Pollination is essential for efficient reproduction in pollinator-dependent crops that rely on the attraction of pollinators to flowers. Especially, floral nectar is considered to be an important factor attracting pollinator like honey bees, but differences among major chestnut species (Castanea
B P Chumpitazi et al.
Alimentary pharmacology & therapeutics, 42(4), 418-427 (2015-06-25)
A low fermentable oligosaccharides, disaccharides, monosaccharides and polyols (FODMAP) diet can ameliorate symptoms in adult irritable bowel syndrome (IBS) within 48 h. To determine the efficacy of a low FODMAP diet in childhood IBS and whether gut microbial composition and/or metabolic
Souhir Marsit et al.
Molecular biology and evolution, 32(7), 1695-1707 (2015-03-10)
Although an increasing number of horizontal gene transfers have been reported in eukaryotes, experimental evidence for their adaptive value is lacking. Here, we report the recent transfer of a 158-kb genomic region between Torulaspora microellipsoides and Saccharomyces cerevisiae wine yeasts
K Shiraga et al.
Carbohydrate research, 406, 46-54 (2015-02-07)
Terahertz time-domain attenuated total reflection measurements of monosaccharide (glucose and fructose) and disaccharide (sucrose and trehalose) solutions from 0.146 M to 1.462 M were performed to evaluate (1) the hydration state and (2) the destructuring effect of saccharide solutes on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service