Skip to Content
Merck
D-(+)-Glucose tested according to Ph. Eur.

D-(+)-Glucose

D-(+)-Glucose tested according to Ph. Eur.
Synonym(s):
Dextrose
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Package Size
Special Grade
Technique