Skip to Content
Merck
All Photos(3)

Key Documents

31494

Sigma-Aldrich

Silver sulfate

puriss. p.a., ACS reagent, ≥99.5%

Synonym(s):

Sulfuric acid disilver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag2SO4
CAS Number:
Molecular Weight:
311.80
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Quality Level

Assay

≥99.5%

form

crystalline

impurities

≤0.02% Insoluble matter and silver chloride
≤0.03% substances not precipitated by HCl

mp

652 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
nitrate (NO3-): ≤10 mg/kg

cation traces

Ca: ≤50 ppm
Cd: ≤50 ppm
Co: ≤50 ppm
Cu: ≤5 ppm
Fe: ≤10 ppm
K: ≤100 ppm
Na: ≤200 ppm
Ni: ≤10 ppm
Pb: ≤10 ppm
Zn: ≤5 ppm

SMILES string

[Ag+].[Ag+].[O-]S([O-])(=O)=O

InChI

1S/2Ag.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

YPNVIBVEFVRZPJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Silver sulfate (Ag2SO4) is a disilver(I) salt of sulfuric acid. It participates in the TiO2-assisted photooxidation of substituted benzyltrimethylsilanes in acetonitrile. Diarylethanes were obtained in 50-74% yields.

Application

Silver sulfate may be employed in the preparation of the following:
  • Iodination reagent in combination with iodine for the synthesis of iododerivatives.
  • Silver sulfate reagent, which is required for the determination of Chemical Oxygen Demand (COD).

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical stability of plasmon-active silver tips for tip-enhanced Raman spectroscopy.
Kalbacova J, et al.
Nanospectroscopy, 1(1), 12-18 (2014)
Ponnusamy Velusamy et al.
Journal of advanced research, 5(1), 19-25 (2015-02-17)
The photocatalytic decoloration of an organic dye, ethyl violet (EV), has been studied in the presence of TiO2 and the addition of β-Cyclodextrin (β-CD) with TiO2 (TiO2-β-CD) under UV-A light irradiation. The different operating parameters like initial concentration of dye
Iodination with Silver Sulfate and Iodine. II. Uridines.
Sy W-W.
Synthetic Communications, 20(21), 3391-3394 (1990)
Iodination of aromatic amines with iodine and silver sulfate.
Sy W-W.
Synthetic Communications, 22(22), 3215-3219 (1992)
Titanium dioxide photocatalysed oxidation of benzyltrimethylsilanes in the presence of silver sulfate.
Baciocchi E, et al.
Journal of the Chemical Society. Chemical Communications, 1, 59-60 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service