Skip to Content
Merck
All Photos(3)

Key Documents

402869

Sigma-Aldrich

Chloramine T trihydrate

ACS reagent, 98%

Synonym(s):

N-Chloro-p-toluenesulfonamide sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2NClNa · 3H2O
CAS Number:
Molecular Weight:
281.69
Beilstein:
3924168
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Assay

98%
98.0-103.0% (ACS specification)

form

powder

reaction suitability

reagent type: oxidant

impurities

<1.5% insolubles (alcohol)

pH

8.0-10.0 (25 °C, 5%)

mp

167-170 °C (lit.)

solubility

H2O: passes test

suitability

passes test for determination of bromide

SMILES string

O.O.O.Cc1ccc(cc1)S(=O)(=O)N([Na])Cl

InChI

1S/C7H7ClNO2S.Na.3H2O/c1-6-2-4-7(5-3-6)12(10,11)9-8;;;;/h2-5H,1H3;;3*1H2/q-1;+1;;;

InChI key

NZYOAGBNMCVQIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloramine T (CAT) trihydrate is a commonly used reagent for organic synthesis especially in heterocyclic chemistry. CAT is an active chlorine compound that shows antimicrobial activity.

Application

Chloramine T trihydrate may be used as an oxidizing agent in the synthesis of 8-chloro-[1,2,4]triazolo[4,3-a]pyrazine derivatives and novel isoxazolines.
It may also be used in the synthesis of the following compounds:
  • Nitrile imines by the oxidative dehydrogenation of N-(nitrobenzyl)-imidazole aldehyde hydrazine.
  • Mono-N-tosylated-1,2-diamines.
  • 3,5-Disubstituted isoxazoles.
  • Fused 3,6-disubstituted triazolothiadiazoles by the oxidative cyclization of N-heteroaryl-substituted hydrazones.
Nitrene source for aziridinations and aminohydroxylations.

Packaging

Capable of oxidative cyclization to produce various heterocycles.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

377.6 °F - closed cup

Flash Point(C)

192 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel (+)-3-Carene Derivatives and Their Application in Asymmetric Synthesis.
Roszkowski P, et al.
Synthesis, 47(04), 569-574 (2015)
Facile one pot synthesis of 8-chloro-[1, 2, 4] triazolo [4, 3-a] pyrazines via oxidative cyclisation using chloramine T.
Mal S, et al.
Tetrahedron Letters, 56(22), 2896-2901 (2015)
Facile Synthesis of 3, 6-Disubstituted-1, 2, 4-triazolo-[3, 4-b]-1, 3, 4-thiadiazoles via Oxidative Cyclization of N-Heteroaryl-Substituted Hydrazones and Their Biological Activity.
Himaja M, et al.
Journal of Heterocyclic Chemistry, 49(4), 823-828 (2012)
Kantam, M. L. et al
Tetrahedron Letters, 44, 9029-9029 (2003)
G D Kishore Kumar et al.
Chemical communications (Cambridge, England), (8), 1026-1027 (2004-04-08)
Environmentally benign HPA is found to be an efficient catalyst for aziridination of olefins in the presence of inexpensive Chloramine-T as a nitrogen source: instantaneous at room temperature, requires only stoichiometric amount of olefin and no allyl amine side product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service