Skip to Content
Merck
All Photos(3)

Key Documents

227927

Sigma-Aldrich

Silver chloride

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgCl
CAS Number:
Molecular Weight:
143.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

1 mmHg ( 912 °C)

Quality Level

product line

ReagentPlus®

Assay

99%

form

powder or chunks

reaction suitability

reagent type: catalyst
core: silver

mp

455 °C (lit.)

SMILES string

Cl[Ag]

InChI

1S/Ag.ClH/h;1H/q+1;/p-1

InChI key

HKZLPVFGJNLROG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Silver chloride (AgCl) is widely employed as a catalyst, photographic material, and ionic semiconductor material. High photosensitivity of AgCl makes it an important photocatalyst.

Application

Silver chloride can be used as a catalyst:
  • In the preparation of 1,2,3-triazoles from alkynes and azides.

Silver chloride is also be used:
  • As a precursor salt to synthesize silver nanoparticles.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Band structure of silver chloride and silver bromide.
Scop PM.
Physical Review, 139(3A), A934-A934 (1965)
Ionic conductivity of silver chloride single crystals.
Corish J and Jacobs PWM.
Journal of Physics and Chemistry of Solids, 33(7), 1799-1818 (1972)
A type of silver chloride electrode suitable for use in dilute solutions.
Brown AS.
Journal of the American Chemical Society, 56(3), 646-647 (1934)
Tomohisa Hayashi et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 28(2), 127-133 (2012-02-11)
Electrochemical detection of sugar-related compounds was conducted using a boron-doped diamond (BDD) electrode as a detector for flow-injection analysis (FIA). Sugar-related compounds oxidize at high applied potentials, for which the BDD electrode is suitable for electrochemical measurements. Conditions for an
Alevtina Neyman et al.
Chemical communications (Cambridge, England), 48(16), 2207-2209 (2012-01-19)
"Out-of-pocket" association of Ag(+) to the tetradentate defect site of mono-vacant Keggin and Wells-Dawson polyoxometalate (POM) cluster-anions is used to direct the formation of water-soluble AgCl nanocubes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service