Skip to Content
Merck
All Photos(3)

Key Documents

F1543

Sigma-Aldrich

Ammonium iron(II) sulfate hexahydrate

BioXtra, ≥98%

Synonym(s):

Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate, Mohr’s salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2Fe(SO4)2 · 6H2O
CAS Number:
Molecular Weight:
392.14
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Quality Level

Assay

≥98%

reaction suitability

reagent type: catalyst
core: iron

impurities

≤0.0005% Phosphorus (P)
≤0.01% Insoluble matter

mp

100 °C (dec.) (lit.)

solubility

dilute sulfuric acid: 1 M

cation traces

Al: ≤0.001%
Ca: ≤0.0005%
Cu: ≤0.005%
K: ≤0.005%
Mg: ≤0.002%
Mn: ≤0.01%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.001%

SMILES string

N.N.O.O.O.O.O.O.[Fe++].OS([O-])(=O)=O.OS([O-])(=O)=O

InChI

1S/Fe.2H3N.2H2O4S.6H2O/c;;;2*1-5(2,3)4;;;;;;/h;2*1H3;2*(H2,1,2,3,4);6*1H2/q+2;;;;;;;;;;/p-2

InChI key

MQLVWQSVRZVNIP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • pH responsive controlled release of anti-cancer hydrophobic drugs from sodium alginate and hydroxyapatite bi-coated iron oxide nanoparticles.: This study explores the use of ammonium iron(II) sulfate hexahydrate in the synthesis of bi-coated iron oxide nanoparticles for pH-responsive controlled drug release. The nanoparticles, coated with sodium alginate and hydroxyapatite, were designed for the controlled release of hydrophobic anti-cancer drugs, demonstrating significant potential for targeted cancer therapy applications (Manatunga et al., 2017).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liangfu Zhou et al.
Free radical research, 54(6), 385-396 (2020-03-19)
Vitamin C and iron are both important nutrients for humans and involved in several physiological processes. The biological activities of vitamin C and iron are based on their abilities to accept or donate electrons. Although vitamin C is well known
Yu-Jung Chang et al.
Nutrition (Burbank, Los Angeles County, Calif.), 21(4), 520-524 (2005-04-07)
This study examined whether iron accumulated in ferritin-producing recombinant microbes is bioavailable to rats with iron deficiency. Rats induced with iron deficiency were treated with iron preparations of ferrous ammonium sulfate, horse spleen ferritin, control yeast, and ferritin-producing recombinant yeast
Ubon Cha'on et al.
Toxicological sciences : an official journal of the Society of Toxicology, 96(1), 194-201 (2006-12-19)
The aim of this study is to identify the biochemical mechanism of phosphine toxicity and resistance, using Caenorhabditis elegans as a model organism. To date, the precise mode of phosphine action is unclear. In this report, we demonstrate the following
Ashish Pathak et al.
Journal of hazardous materials, 171(1-3), 273-278 (2009-07-10)
The potential of indigenous iron-oxidizing microorganisms enriched at initial neutral pH of the sewage sludge for bioleaching of heavy metals was investigated at initial neutral pH of the sludge using ammonium ferrous sulfate (FAS) and ferrous sulfate (FS) as an
S Brindha et al.
Physics in medicine and biology, 47(11), N153-N158 (2002-07-11)
Radiation dosimetry deals with the determination of absorbed dose to the medium exposed to ionizing radiation. Chemical dosimetry depends on oxidation or reduction of chemicals by ionizing radiation. A ferrous ammonium sulfate benzoic acid xyelenol orange (FBX) dosimeter based on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service