Skip to Content
Merck
All Photos(1)

Key Documents

V0033000

Valproic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Propylpentanoic acid, Valproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)2CHCO2H
CAS Number:
Molecular Weight:
144.21
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference Standard

API family

valproic acid

manufacturer/tradename

EDQM

refractive index

n20/D 1.425 (lit.)

bp

220 °C (lit.)

density

0.9 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCC(C(O)=O)CCC

InChI

1S/C8H16O2/c1-3-5-7(6-4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)

InChI key

NIJJYAXOARWZEE-UHFFFAOYSA-N

Gene Information

human ... ALDH5A1(7915)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Valproic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Anticonvulsant that also has efficacy as a mood stabilizer in bipolar disorder

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1A - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

231.8 °F - closed cup

Flash Point(C)

111 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mattias Linde et al.
The Cochrane database of systematic reviews, 6(6), CD010611-CD010611 (2013-06-26)
Some antiepileptic drugs but not others are useful in clinical practice for the prophylaxis of migraine. This might be explained by the variety of actions of these drugs in the central nervous system. The present review is part of an
Chi-Tso Chiu et al.
Pharmacological reviews, 65(1), 105-142 (2013-01-10)
The mood stabilizers lithium and valproic acid (VPA) are traditionally used to treat bipolar disorder (BD), a severe mental illness arising from complex interactions between genes and environment that drive deficits in cellular plasticity and resiliency. The therapeutic potential of
Radu M Nanau et al.
Clinical biochemistry, 46(15), 1323-1338 (2013-06-25)
Valproic acid is a widely-used first-generation antiepileptic drug, prescribed predominantly in epilepsy and psychiatric disorders. VPA has good efficacy and pharmacoeconomic profiles, as well as a relatively favorable safety profile. However, adverse drug reactions have been reported in relation with
Taylor Chomiak et al.
Neurotoxicology and teratology, 36, 57-66 (2012-09-13)
Autism spectrum disorder (ASD) is a behaviourally defined brain disorder affecting approximately 1 in 88 children. Many pathological studies have shown that ASD is frequently associated with grey and white matter changes that can be described by their deviations from
Alexa P Vitins et al.
Archives of toxicology, 88(8), 1573-1588 (2014-02-19)
Liver injury is the leading cause of drug-induced toxicity. For the evaluation of a chemical compound to induce toxicity, in this case steatosis or fatty liver, it is imperative to identify markers reflective of mechanisms and processes induced upon exposure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service