Skip to Content
Merck
All Photos(1)

Key Documents

PHR1061

Supelco

Valproic acid

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-Propylpentanoic acid, 2-Propylvaleric acid, Valproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)2CHCO2H
CAS Number:
Molecular Weight:
144.21
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. V0033000
traceable to USP 1708707

API family

valproic acid

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.425 (lit.)

bp

220 °C (lit.)

density

0.9 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCCC(C(O)=O)CCC

InChI

1S/C8H16O2/c1-3-5-7(6-4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)

InChI key

NIJJYAXOARWZEE-UHFFFAOYSA-N

Gene Information

human ... ALDH5A1(7915)

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.
It is widely used in the treatment of bipolar disorder, migraine, depression, epilepsy and schizophrenia.[1]
Valproic acid is a branched short chain fatty acid, which finds applications as a antiepileptic drug.[2][3]

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Valproic acid may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations using gas chromatography technique.[1]

Biochem/physiol Actions

Anticonvulsant that also has efficacy as a mood stabilizer in bipolar disorder

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB3712 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1A - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

231.8 °F - closed cup

Flash Point(C)

111 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of microsomal Phenobarbital metabolism by valproic acid
Kapetanovic MI and Kupferberg JH
Biochemical Pharmacology, 30(11), 1361-1363 (1981)
Histone deacetylase is a direct target of valproic acid, a potent anticonvulsant, mood stabilizer, and teratogen
Phiel JC, et al.
The Journal of Biological Chemistry, 276(39), 36734-36741 (2001)
Headspace solid-phase microextraction-gas chromatography method for the determination of valproic acid in human serum, and formulations using hollow-fiber coated wire
Farajzadeh AM, et al.
Analytical Sciences, 25(7), 875-879 (2009)
Jakob Christensen et al.
JAMA, 309(16), 1696-1703 (2013-04-25)
Valproate is used for the treatment of epilepsy and other neuropsychological disorders and may be the only treatment option for women of childbearing potential. However, prenatal exposure to valproate may increase the risk of autism. To determine whether prenatal exposure
Xiaolei Yin et al.
Nature methods, 11(1), 106-112 (2013-12-03)
Although Lgr5(+) intestinal stem cells have been expanded in vitro as organoids, homogeneous culture of these cells has not been possible thus far. Here we show that two small molecules, CHIR99021 and valproic acid, synergistically maintain self-renewal of mouse Lgr5(+)

Questions

1–2 of 2 Questions  
  1. Could you please provide me with information regarding the solvent used to dissolve this product and the final concentration of valproic acid in 1G pack (as g/ml) Additionally, I would like to know if valproic acid available for purchase in powder form

    1 answer
    1. In the pure form, Valproic acid exists as a neat liquid, the concentration will be considered 100%. The density of the pure form is 0.905 g/mL at 20 °C. The solubility is not tested, however literature shows that it is only slightly soluble in water - 1.3 - 2.0 mg/mL. It is freely soluble in organic solvents, acids and bases. The salt forms are available as powders. Please see the links below to review additional solubility information available at PubChem, as well as a link to the powder form of this product.

      PubChem - see section 3:
      https://pubchem.ncbi.nlm.nih.gov/compound/Valproic-Acid#section=Solubility

      Valproic acid sodium salt:
      https://www.sigmaaldrich.com/US/en/product/sigma/p4543

      Helpful?

  2. Is it necessary to dissolve in any solvent (like water or serum physiologic) for administration to rats intraperitoneally. Or can it be applied directly?

    1 answer
    1. Valproic acid is reportedly water soluble at 50 mg/mL. However, this product has not been tested for in vivo or in vitro use. This product is not sterile, suitability would have to be determined by the end-user.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service